Merck
  • Isolation and characterization of a novel acid degradation impurity of Amlodipine Besylate using Q-TOF, NMR, IR and single crystal X-ray.
Isolation and characterization of a novel acid degradation impurity of Amlodipine Besylate using Q-TOF, NMR, IR and single crystal X-ray.
Journal of pharmaceutical and biomedical analysis (2014-07-30)
Ravi Rapolu, Ch Krishnam Raju, Kolupula Srinivas, Atul Awasthi, Sameer G Navalgund, Koduru V Surendranath
ZUSAMMENFASSUNG
Forced degradation of Amlodipine Besylate (AMD) in acidic condition gave rise to a potential unknown impurity. This unknown acid degradation product (ADP) was evaluated using a new-reverse-phase high performance liquid chromatography (HPLC), where it was eluted at 1.24 relative retention time to AMD peak. ADP was isolated using preparative HPLC from degradation mixture. Later, structure of ADP was elucidated using high resolution MS, multidimensional NMR and FTIR spectroscopic techniques, and characterized as ethyl-6-(2-chlorophenyl)-8-methyl-3,4,6,7-tetrahydro-2H-benzo[b][1,4]oxazine-5-carboxylate. The presence of ADP recemic mixture was confirmed by polarimeter and chiral HPLC. Given the complexity associated with ADP generation, single crystal X-ray crystallography technique was used to confirm proposed structure. In addition, reaction mechanism was postulated and confirmed using computational chemistry. To our knowledge, it is a novel impurity and not reported elsewhere.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Ammoniumacetat, ACS reagent, ≥97%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ammoniumacetat, ≥99.99% trace metals basis
Supelco
Ammoniumacetat, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%