Merck
  • A cyclic peptide accelerates the loading of peptide antigens in major histocompatibility complex class II molecules.
A cyclic peptide accelerates the loading of peptide antigens in major histocompatibility complex class II molecules.
Biochemical and biophysical research communications (2014-12-20)
Saifullah Afridi, Farzana Shaheen, Olaf Roetzschke, Zafar Ali Shah, Syed Comail Abbas, Rizwana Siraj, Talat Makhmoor
ZUSAMMENFASSUNG
Major histocompatibility complex (MHC)-loading enhancers (MLE) have recently attracted attention because of their ability to enhance the efficacy of peptide immunotherapeutics. As small molecular weight compounds, they influence the loading of peptides in MHC molecules by converting them from a non-receptive to a receptive state. Herein, we report a 14-mer cyclic peptide 1 (CP-1) as a new class of MLE-peptide. This peptide was used to investigate its loading on human leukocyte antigen (HLA)-DR molecules. It was found that CP-1 strongly accelerates peptide-loading on both soluble and cell surface HLA-DR molecules in a dose-dependent manner. The effect was evident for all subsets of HLA-DR tested, including HLA-DRB1*1501, indicating that it acts independently of P1-pocket size, which is the canonical MLE-binding site. Importantly, increased peptide-loading by CP-1 was correlated with improved CD4(+) T cell responses in vitro, while propidium iodide staining indicated low peptide-induced cytotoxicity. Thus, this study revealed a new class of peptide-based enhancers that catalyze peptide-loading by allosteric interactions with MHC molecules. Because of its low cellular cytotoxicity and high MLE activity, it may be useful in stimulating antigen-specific T cell responses for therapeutic purposes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumpyruvat, BioXtra, ≥99%
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O