Merck
Intact human amniotic membrane differentiated towards the chondrogenic lineage.
Cell and tissue banking (2014-05-16)
Andrea Lindenmair, Sylvia Nürnberger, Guido Stadler, Alexandra Meinl, Christa Hackl, Johann Eibl, Christian Gabriel, Simone Hennerbichler, Heinz Redl, Susanne Wolbank
ZUSAMMENFASSUNG
Human amniotic membrane (hAM) represents a tissue that is well established as biomaterial in the clinics with potential for new applications in regenerative medicine. For tissue engineering (TE) strategies, cells are usually combined with inductive factors and a carrier substrate. We have previously recognized that hAM represents a natural, preformed sheet including highly potent stem cells. In the present approach for cartilage regeneration we have induced chondrogenesis in hAM in vitro. For this, hAM biopsies were cultured for up to 56 days under chondrogenic conditions. The induced hAM was characterized for remaining viability, glycosaminoglycan (GAG) accumulation using histochemical analysis, and a quantitative assay. Collagen I, II and X was immunohistochemically determined and cartilage-specific mRNA expression of (sex determining region Y-) box 9, cartilage oligomeric matrix protein (COMP), aggrecan (AGC1), versican (CSPG2), COL1A1, COL9A2, melanoma inhibitory activity (MIA), and cartilage-linking protein 1 (CRTL1) analyzed by quantitative real-time polymerase chain reaction. Human AM was successfully induced to accumulate GAG, as demonstrated by Alcianblue staining and a significant (p < 0.001) increase of GAG/viability under chondrogenic conditions peaking in a 29.9 ± 0.9-fold induction on day 56. Further, upon chondrogenic induction collagen II positive areas were identified within histological sections and cartilage-specific markers including COMP, AGC1, CSPG2, COL1A1, COL9A2, MIA, and CRTL1 were found upregulated at mRNA level. This is the first study, demonstrating that upon in vitro induction viable human amnion expresses cartilage-specific markers and accumulates GAGs within the biomatrix. This is a promising first step towards a potential use of living hAM for cartilage TE.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Linoleic acid, ≥98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Cystein -hydrochlorid, anhydrous, ≥98% (TLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Cystein -hydrochlorid, anhydrous, from non-animal source, BioReagent, suitable for cell culture, ≥98.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Cystein -hydrochlorid, anhydrous, ≥99.0% (RT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Linolsäure, technical, 58-74% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Linolsäure, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.