Merck
  • Serum fatty acid profiles and potential biomarkers of ankylosing spondylitis determined by gas chromatography-mass spectrometry and multivariate statistical analysis.
Serum fatty acid profiles and potential biomarkers of ankylosing spondylitis determined by gas chromatography-mass spectrometry and multivariate statistical analysis.
Biomedical chromatography : BMC (2014-09-17)
Rui Chen, Su Han, Daming Dong, Yansong Wang, Qingpeng Liu, Wei Xie, Mi Li, Meng Yao
ZUSAMMENFASSUNG
Ankylosing spondylitis (AS) is a common chronic inflammatory rheumatic disease. Early and accurate detection is essential for effective disease treatment. Recently, research has focused on genomics and proteomics. However, the associated metabolic variations, especially fatty acid profiles, have been poorly discussed. In this study, the gas chromatography-mass spectrometry (GC-MS) approach and multivariate statistical analysis were used to investigate the metabolic profiles of serum free fatty acids (FFAs) and esterified fatty acids (EFAs) in AS patients. The results showed that significant differences in most of the FFA (C12:0, C16:0, C16:1, C18:3, C20:4, C20:5, C22:5 and C22:6) and EFA (C12:0, C16:1, C18:0, C18:1, C18:2, C18:3, C20:4 and C22:6) concentrations were found between the AS patients and healthy controls (p < 0.05). Principal component analysis and partial least squares discriminant analysis were performed to classify the AS patients and controls. Additionally, FFAs C20:4, C12:0, C18:3 and EFAs C22:6, C12:0 were confirmed as potential biomarkers to identify AS patients and healthy controls. The present study highlights that differences in the serum FFA and EFA profiles of AS patients reflect the metabolic disorder. Moreover, FFA and EFA biomarkers appear to have clinical applications for the screening and diagnosis of AS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Sigma-Aldrich
Palmitinsäure, ≥99%
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Linolensäure, ≥99%
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Stearinsäure, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Oleinsäure, BioReagent, suitable for cell culture
Sigma-Aldrich
Laurinsäure, ≥98%, FCC, FG
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Heptadecansäure, ≥98%
Sigma-Aldrich
Palmitoleinsäure, ≥98.5% (GC), liquid
Sigma-Aldrich
Stearinsäure, reagent grade, 95%
Sigma-Aldrich
Palmitinsäure, BioXtra, ≥99%
Sigma-Aldrich
Myristinsäure, Sigma Grade, ≥99%
Sigma-Aldrich
Arachidonsäure, >95.0% (GC)
Sigma-Aldrich
Oleinsäure, natural, FCC
Sigma-Aldrich
Oleinsäure, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Palmitinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Stearinsäure, ≥95%, FCC, FG
Sigma-Aldrich
Arachidonsäure, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Laurinsäure, 98%
Sigma-Aldrich
Palmitinsäure, ≥98%, FCC, FG
USP
Stearinsäure, United States Pharmacopeia (USP) Reference Standard
USP
Palmitinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Linolsäure, technical, 58-74% (GC)
Supelco
Stearinsäure, analytical standard
Supelco
Oleinsäure, analytical standard
Supelco
Linolsäure, analytical standard
Supelco
Palmitinsäure, analytical standard
Sigma-Aldrich
Laurinsäure, natural, ≥98%, FCC, FG