Merck
  • Switch-loop flexibility affects transport of large drugs by the promiscuous AcrB multidrug efflux transporter.
Switch-loop flexibility affects transport of large drugs by the promiscuous AcrB multidrug efflux transporter.
Antimicrobial agents and chemotherapy (2014-06-11)
Hi-jea Cha, Reinke T Müller, Klaas M Pos
ZUSAMMENFASSUNG
Multidrug efflux transporters recognize a variety of structurally unrelated compounds for which the molecular basis is poorly understood. For the resistance nodulation and cell division (RND) inner membrane component AcrB of the AcrAB-TolC multidrug efflux system from Escherichia coli, drug binding occurs at the access and deep binding pockets. These two binding areas are separated by an 11-amino-acid-residue-containing switch loop whose conformational flexibility is speculated to be essential for drug binding and transport. A G616N substitution in the switch loop has a distinct and local effect on the orientation of the loop and on the ability to transport larger drugs. Here, we report a distinct phenotypical pattern of drug recognition and transport for the G616N variant, indicating that drug substrates with minimal projection areas of >70 Å(2) are less well transported than other substrates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Chloramphenicol, VETRANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.