Merck
Serum vitamin D, vitamin D binding protein, and lung cancer survival.
Lung cancer (Amsterdam, Netherlands) (2014-12-03)
Gabriella M Anic, Stephanie J Weinstein, Alison M Mondul, Satu Männistö, Demetrius Albanes
ZUSAMMENFASSUNG
Vitamin D may prolong cancer survival by inhibiting tumor progression and metastasis, however, there are limited epidemiologic studies regarding the association between circulating 25-hydroxyvitamin D (25(OH)D) and lung cancer survival. The aim of this study was to examine the relationship between serum 25(OH)D and lung cancer specific survival and to evaluate whether vitamin D binding protein (DBP) concentration modified this association. 25(OH)D and DBP were measured in fasting serum samples from 500 male lung cancer cases in the Alpha-Tocopherol, Beta-Carotene Cancer Prevention Study. Cox proportional hazards regression was used to estimate hazard ratios (HRs) and 95% confidence intervals (CI) for lung cancer related death according to quartiles of season-specific 25(OH)D, DBP, and the molar ratio of 25(OH)D:DBP, a proxy for free circulating 25(OH)D. Comparing highest to lowest quartiles, serum 25(OH)D (HR=1.18; 95% CI: 0.89-1.56) and DBP (HR=0.95; 95% CI: 0.71-1.26) were not associated with lung cancer survival and DBP concentration did not modify the association with 25(OH)D (p for interaction=0.56). There was suggestion of an association between higher serum 25(OH)D and better survival from adenocarcinoma (HR=0.64; 95% CI: 0.17-2.45) and small cell carcinoma (HR=0.55; 95% CI: 0.21-1.45), but these estimates were based on a relatively small number of cases. Serum 25(OH)D was not associated with overall lung cancer survival regardless of DBP concentration, however, these findings should be examined in other studies that include women and subjects with higher 25(OH)D levels.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Natriumphosphat dibasisch Dodecahydrat, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Phthalsäuredibutylester, 99%
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Supelco
α-Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sigma-Aldrich
β-Carotin, synthetic, ≥95% (HPLC), crystalline
Supelco
(±)-α-Tocopherol, analytical standard
Sigma-Aldrich
β-Carotin, synthetic, ≥93% (UV), powder
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Natriumphosphat dibasisch Dodecahydrat, BioXtra, ≥99.0% (T)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Calcium, granular, 99%
Supelco
β-Carotin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumphosphat dibasisch Dodecahydrat, tested according to Ph. Eur.