Merck
G-CSF displays restricted ability to promote Sca-1(+) cardiac stem cell proliferation in vitro.
Molecular and cellular biochemistry (2014-08-28)
Haijian Luo, Giulio Bassi, Maddalena Tessari, Zhenyu Yang, Giuseppe Faggian
ZUSAMMENFASSUNG
Granulocyte colony-stimulating factor (G-CSF) is a controversial chemical in cardiac cell therapy. Myocardial homing of mobilized bone marrow-derived cells is thought to play a critical role in observed G-CSF-induced cardiac repair; meanwhile, the activation of proliferative potential of cardiac stem cells (CSCs) residing in the heart is a significant challenge. The present study aims to investigate whether G-CSF receptor is expressed in adult resident Sca-1(+) CSCs and determine the effect of G-CSF treatment on the proliferation of CSCs. For cardiac cells isolation, 12-week-old male C57BL/6 mice were anesthetized in a chamber containing 2.5% isoflurane in oxygen, euthanized by CO2 inhalation and then sacrificed by cervical dislocation. Magnetic-activated cell sorting was employed to acquire highly purified Sca-1(+) CSCs. We found that G-CSF receptor was expressed in adult resident Sca-1(+) CSCs by immunofluorescence staining and Western blotting. Exposure of Sca-1(+) cells to G-CSF in the culture medium for 72 h induced time-dependent but self-limiting cell cycle acceleration with a restricted effect on the CSC proliferation. As a result, it has provided a new insight to focus on the association between cardiac G-CSF therapy and adult resident stem cell activation. It may suggest gaining a deeper insight into the mechanisms of the interaction between CSCs and G-CSF to develop a synergistic strategy based on resident stem cell and G-CSF therapy for heart disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Natriumcitrat, dreibasisch Dihydrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Deoxycholsäure Natriumsalz, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Deoxycholsäure Natriumsalz, ≥97% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Natriumcitrat-Dihydrat, ≥99%, FG
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Trinatriumcitrat Dihydrat, meets USP testing specifications
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material