Merck
  • The role of total and cartilage-specific estrogen receptor alpha expression for the ameliorating effect of estrogen treatment on arthritis.
The role of total and cartilage-specific estrogen receptor alpha expression for the ameliorating effect of estrogen treatment on arthritis.
Arthritis research & therapy (2014-07-17)
Cecilia Engdahl, Anna E Börjesson, Huamei F Forsman, Annica Andersson, Alexandra Stubelius, Andree Krust, Pierre Chambon, Ulrika Islander, Claes Ohlsson, Hans Carlsten, Marie K Lagerquist
ZUSAMMENFASSUNG
Estrogen (E2) delays onset and decreases severity of experimental arthritis. The aim of this study was to investigate the importance of total estrogen receptor alpha (ERα) expression and cartilage-specific ERα expression in genetically modified mice for the ameliorating effect of estrogen treatment in experimental arthritis. Mice with total (total ERα-/-) or cartilage-specific (Col2α1-ERα-/-) inactivation of ERα and wild-type (WT) littermates were ovariectomized, treated with E2 or placebo, and induced with antigen-induced arthritis (AIA). At termination, knees were collected for histology, synovial and splenic cells were investigated by using flow cytometry, and splenic cells were subjected to a T-cell proliferation assay. E2 decreased synovitis and joint destruction in WT mice. Amelioration of arthritis was associated with decreased frequencies of inflammatory cells in synovial tissue and decreased splenic T-cell proliferation. E2 did not affect synovitis or joint destruction in total ERα-/- mice. In Col2α1-ERα-/- mice, E2 protected against joint destruction to a similar extent as in WT mice. In contrast, E2 did not significantly ameliorate synovitis in Col2α1-ERα-/- mice. Treatment with E2 ameliorates both synovitis and joint destruction in ovariectomized mice with AIA via ERα. This decreased severity in arthritis is associated with decreased synovial inflammatory cell frequencies and reduced splenic T-cell proliferation. ERα expression in cartilage is not required for estrogenic amelioration of joint destruction. However, our data indicate that ERα expression in cartilage is involved in estrogenic effects on synovitis, suggesting different mechanisms for the amelioration of joint destruction and synovitis by E2.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
SAFC
L-Glutamin
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Aphidicolin, ≥98% (HPLC), powder
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Carprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Carprofen, VETRANAL®, analytical standard
Isofluran, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Aphidicolin, analytical standard
Carprofen for veterinary use, European Pharmacopoeia (EP) Reference Standard
Carprofen für die Systemeignung, European Pharmacopoeia (EP) Reference Standard