Merck
  • Profiling of stable isotope enrichment in specialized metabolites using liquid chromatography and multiplexed nonselective collision-induced dissociation.
Profiling of stable isotope enrichment in specialized metabolites using liquid chromatography and multiplexed nonselective collision-induced dissociation.
Analytical chemistry (2014-10-10)
Zhenzhen Wang, A Daniel Jones
ZUSAMMENFASSUNG
Engineering of specialized metabolites in plants, microbes, and other organisms is hindered by significant knowledge gaps about metabolic pathways responsible for metabolite accumulation and degradation. While isotopic tracers have provided important information about metabolic fluxes in central metabolism, limitations of mass spectrometric strategies for quantifying stable isotope incorporation into both intact metabolites and specific substructures have slowed extension of these techniques to large specialized metabolites. This report describes the application of electrospray ionization with data-independent multiplexed nonselective collision induced dissociation (CID) on a time-of-flight mass spectrometer. This strategy yields quasi-simultaneous collection, on the chromatographic time scale, of mass spectra with different degrees of fragment ion formation without biases introduced by precursor mass selection or selective ion activation and provides measurements of stable isotope enrichments in intact metabolites and individual substructures. The utility and precision of these analyses is demonstrated by labeling acylsugar metabolites in glandular trichomes of tomato (Solanum lycopersicum) using (13)CO2 and analyzing (13)C enrichments in acylsugar specialized metabolites. The high precision and avoidance of mass bias provide a promising tool for extending metabolic flux analyses to complex specialized metabolites in a wide range of organisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Schwefelsäure, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard