Merck
  • Identification of prometon, deisopropylprometon, and hydroxyprometon in groundwater by high resolution liquid chromatography/mass spectrometry.
Identification of prometon, deisopropylprometon, and hydroxyprometon in groundwater by high resolution liquid chromatography/mass spectrometry.
The Science of the total environment (2014-08-26)
C Fernández-Ramos, Imma Ferrer, Karl Mauch, D Satinsky, E Michael Thurman
ZUSAMMENFASSUNG
Prometon, a major soil sterilant, and its main transformation products, deisopropylprometon (N(2)-isopropyl-6-methoxy-1,3,5-triazine-2,4-diamine) and hydroxyprometon (4,6-bis(isopropylamino)-1,3,5-triazin-2-ol), were identified as the dominant triazine herbicides in groundwater samples from 51 locations in Colorado, USA, over a two-year time period. They were concentrated from water by solid phase extraction and detected using an ultrahigh pressure, liquid chromatography-quadrupole time of flight tandem mass spectrometry (UHPLC/QTOF-MS). The transformation products, deisopropylprometon and hydroxyprometon, were confirmed using MS-MS experiments. An original strategy was applied to form the degradation standards for deisopropylprometon and hydroxyprometon, which consisted of photo-degradation of prometon followed by MS-MS analysis. The concentration of prometon ranged from the detection limit of 3 ng·L(-1) to 87 ng·L(-1), hydroxyprometon ranged up to 50 ng·L(-1), and deisopropylprometon up to 100 ng·L(-1), with a frequency of detection of 80%, which was greater than the other triazines detected in the groundwater samples. A new ratio is proposed for prometon degradation called the "deisopropylprometon to prometon ratio" or the DIP ratio, as an indicator of prometon residence time in groundwater. Furthermore, these data suggest that prometon is more of an issue for groundwater contamination in urban areas rather than agricultural areas.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Carbamazepin, powder
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Supelco
Carbamazepin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Carbamazepin, analytical standard
USP
Carbamazepin, United States Pharmacopeia (USP) Reference Standard
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard