Merck
Anti-biofilm activity of Marula - a study with the standardized bark extract.
Journal of ethnopharmacology (2014-04-20)
Ratul Sarkar, Sushil K Chaudhary, Amrita Sharma, Kirendra K Yadav, Neelesh K Nema, Mamello Sekhoacha, Sanmoy Karmakar, Fernão C Braga, Motlalepula Gilbert Matsabisa, Pulok K Mukherjee, Tuhinadri Sen
ZUSAMMENFASSUNG
Marula (Sclerocarya birrea; family - Anacardiaceae) is an African plant, which enjoys wide socio-economic importance particularly in southern part of Africa. The fruits are consumed as food and also as alcoholic beverage (cream liquor). In different parts of Africa, the decoction of the bark is traditionally used for the treatment of dysentery, diarrhoea, and various other infectious conditions. The aim of the study was to investigate the anti-biofilm properties of the methanol extract of Marula bark (stem bark of Sclerocarya birrea), with a view towards combating the emergence of antimicrobial resistance often associated with bacterial biofilms. The standardized methanol extract was initially tested for its antimicrobial property. The crystal violet assay was used for evaluating anti-biofilm (biofilm formation by Pseudomonas aeuginosa) activity. Further in order to study the mechanism of anti-biofilm activity, the same was evaluated for understanding its role on various quorums sensing mediated phenomenon (swarming motility assay, protease and pyoverdin assay) that are known to be associated with the formation of biofilms and pathogenicity. The methanol extract showed no inhibition of bacterial growth up to a concentration of 200 µg/ml. Interestingly, the sample produced anti-biofilm activity (around 75% decrease; 100 µg/ml) at sub-lethal concentration. Further it also significantly reduced the QS mediated swarming motility. The release of various virulent factors (protease and pyoverdin) was found to be lowered when pre-treated with the extract. The present study illustrates the anti-biofilm property Sclerocarya birrea. The standardized extract significantly disrupted the quorum sensing mediated production of biofilm formation and also inhibited swarming ability of the cells. The extract displayed a regulatory role on the secretion of protease and pyoverdin, two QS dependent pathogenic factors found in Pseudomonas aeruginosa. This study also validates the ethnobotanical use of Marula.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Trizma® Base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Tris(hydroxymethyl)aminomethan, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Tromethamin, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Trizma® Base, BioUltra, Molecular Biology, ≥99.8% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Trizma® Base, ≥99.0% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Trizma® Base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Trizma® Base, ≥99.9% (titration), crystalline
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Trizma® Base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Tromethamin, pharmaceutical secondary standard, certified reference material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Trizma® Base, 99.7% (T), puriss. p.a.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Trizma® Base, reference material for titrimetry, certified by BAM, >99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Tromethamin, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Trometamol, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
Tromethamin
Preise und Verfügbarkeit sind derzeit nicht verfügbar.