Merck
  • Heterologous expression of AtMYB12 in kale (Brassica oleracea var. acephala) leads to high flavonol accumulation.
Heterologous expression of AtMYB12 in kale (Brassica oleracea var. acephala) leads to high flavonol accumulation.
Plant cell reports (2014-05-06)
Mika Lännenpää
ZUSAMMENFASSUNG
Overexpression of Arabidopsis AtMYB12 transcription factor greatly increases the total phenolic and flavonol content in transgenic kale leaves. Flavonoids are a diverse group of plant secondary metabolites exhibiting a number of health-promoting effects. There has been a growing interest to develop biotechnological methods for the enhanced production of flavonoids in crop plants. AtMYB12 is an Arabidopsis transcription factor which specifically activates flavonol synthesis and its overexpression has led to increased flavonol accumulation in several transgenic plants. In the present study, AtMYB12 was overexpressed in a commercial cultivar of kale and the transgenic plants were tested both in in vitro and in semi-field conditions in cages under natural light. Using this method, a severalfold increase in both total phenolics content and flavonol accumulation was achieved. This study provides a reliable and efficient transformation protocol for kale and suggests the potential of this flavonol-enriched vegetable for the production of kaempferol.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol -Lösung, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Verflüssigtes Phenol, ≥89.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, Molecular Biology
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Supelco
Phenol -Lösung, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol -Lösung, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, natural, 97%, FG
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Phenol -Lösung, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard