Merck
Are 1,4- and 1,5-disubstituted 1,2,3-triazoles good pharmacophoric groups?
ChemMedChem (2014-08-01)
Alberto Massarotti, Silvio Aprile, Valentina Mercalli, Erika Del Grosso, Giorgio Grosa, Giovanni Sorba, Gian Cesare Tron
ZUSAMMENFASSUNG
Over the last decade, 1,2,3-triazoles have received increasing attention in medicinal chemistry thanks to the discovery of the highly useful and widely applicable 1,3-dipolar cycloaddition reaction between azides and alkynes (click chemistry) catalyzed by copper salts and ruthenium complexes. After a decade of medicinal chemistry research on 1,2,3-triazoles, we feel that the time is ripe to demonstrate the real ability of this heterocycle to participate in important and pivotal binding interactions with biological targets while maintaining a good pharmacokinetic profile. In this study, we retrieved and analyzed X-ray crystal structures of complexes between 1,2,3-triazoles and either proteins or DNA to understand the pharmacophoric role of the triazole. Furthermore, the metabolic stability, the capacity to inhibit cytochromes, and the contribution of 1,2,3-triazoles to the overall aqueous solubility of compounds containing them have been analyzed. This information should furnish fresh insight for medicinal chemists in the design of novel bioactive molecules that contain the triazole nucleus.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Acetylaceton, ReagentPlus®, ≥99%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Trichloressigsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloressigsäure -Lösung, 6.1 N
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Uridin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Uridin, ≥99%
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Trichloressigsäure, BioUltra, ≥99.5% (T)