Merck
  • In-situ forming gel-like depot of a polyaspartamide-polylactide copolymer for once a week administration of sulpiride.
In-situ forming gel-like depot of a polyaspartamide-polylactide copolymer for once a week administration of sulpiride.
The Journal of pharmacy and pharmacology (2014-09-23)
Calogero Fiorica, Fabio Salvatore Palumbo, Giovanna Pitarresi, Mario Giorgi, Filippo Calascibetta, Gaetano Giammona
ZUSAMMENFASSUNG
An in-situ forming gel-like depot, prepared by using an appropriate polyaspartamide-polylactide graft copolymer, has been employed to release in a sustained way sulpiride. α,β-poly(N-2-hydroxyethyl)-D,L-aspartamide-g-polylactic acid (PHEA-g-PLA) has been used as a polymer component. Its physicochemical properties make possible to dissolve it in N-methyl-2-pyrrolidone, with the obtainment of a solution able to form a gel-like depot once injected into a physiological medium. Cell compatibility of PHEA-g-PLA depot has been investigated, using murine dermal fibroblasts as cell model. 3-(4,5-Dimethylthiazol-2-yl)-5-(3-carboxymethoxyphenyl)-2-(4-sulfophenyl)-2H-tetrazolium, inner salt assay and fluorescence microscopy have been employed to evaluate cell viability and morphology after contact with PHEA-g-PLA depot. Pharmacokinetic parameters of sulpiride released from depot have been determined following subcutaneous administration to rabbits and compared with corresponding parameters following administration of free sulpiride solution. It has been demonstrated that the system does not affect significantly the viability of fibroblasts and is able to sustain the release of sulpiride until a week, with a burst effect dependent on the initial weight ratio polymer/drug. In-vivo release profiles and pharmacokinetic parameters suggest that PHEA-g-PLA depot could have interesting clinical applications for a once a week administration of poorly soluble drugs to humans or animals.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
1-Methyl-2-pyrrolidon, ACS reagent, ≥99.0%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
1-Methyl-2-pyrrolidon, ReagentPlus®, 99%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
1-Methyl-2-pyrrolidon, anhydrous, 99.5%
Sigma-Aldrich
1-Methyl-2-pyrrolidon, suitable for HPLC, ≥99%
Sigma-Aldrich
N-Hydroxysuccinimid, 98%
Sigma-Aldrich
1-Methyl-2-pyrrolidon, biotech. grade, ≥99.7%
Sigma-Aldrich
Diethylamin, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
N,N-Diethylanilin, ≥99%
Sigma-Aldrich
Diethylamin, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Diethylamin, purified by redistillation, 99.5%
Supelco
Dimethylsulfoxid, analytical standard
Sigma-Aldrich
N-Hydroxysuccinimid, purum, ≥97.0% (T)
Supelco
1-Methyl-2-pyrrolidon, analytical standard
Sigma-Aldrich
(±)-Sulpirid
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)