Merck
Tamoxifen induces the development of hernia in mice by activating MMP-2 and MMP-13 expression.
Biochimica et biophysica acta (2015-02-24)
Xingzhe Ma, Ying Liu, Qixue Wang, Yuanli Chen, Mengyang Liu, Xiaoju Li, Rong Xiang, Yuquan Wei, Yajun Duan, Jihong Han
ZUSAMMENFASSUNG
Hernia is a disease with defects in collagen synthesis/metabolism. However, the underlying mechanisms for hernia formation have not been fully defined. Tamoxifen is a selective estrogen receptor modulator and used for patients with breast cancer. Tamoxifen also has pleiotropic and side effects. Herein, we report that tamoxifen treatment resulted in an appearance of a large bulge in the low abdomen between the hind legs in male but not in female mice. The autopsy demonstrated that the low abdominal wall was broken and a large amount of intestine herniated out of the abdominal cavity. Histological analysis indicated that tamoxifen caused structural abnormalities in the low abdominal wall which were associated with decreased type II collagen content. Furthermore, we determined increased matrix metalloproteinase-2 (MMP-2) and MMP-13 expression in the tissue. In vitro, tamoxifen induced MMP-2 and MMP-13 expression in fibroblasts. The promoter activity analysis and ChIP assay demonstrate that induction of MMP-13 expression was associated with activation of JNK-AP-1 and ERK1/2 signaling pathways while induction of MMP-2 expression was related to activation of the ERK1/2 signaling pathway. Taken together, our study establishes a novel murine hernia model, defines a severe side effect of tamoxifen, and suggests a caution to male patients receiving tamoxifen treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tamoxifen, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Hämatoxylin
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Tamoxifen, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumtrichloracetat, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.