Merck
  • Genotoxicity assessment of melamine in the in vivo Pig-a mutation assay and in a standard battery of assays.
Genotoxicity assessment of melamine in the in vivo Pig-a mutation assay and in a standard battery of assays.
Mutation research. Genetic toxicology and environmental mutagenesis (2015-03-03)
Honggang Tu, Ming Zhang, Changhui Zhou, Zheng Wang, Pengcheng Huang, Hongmei Ou, Yan Chang
ZUSAMMENFASSUNG
The genotoxicity of melamine was evaluated with the combined Pig-a mutation/micronucleus assay, the bacterial reverse mutation assay, and the in vitro cytokinesis-block micronucleus assay (CBMN). Five groups of six- to eight-week-old male Sprague-Dawley (SD) rats were given three daily doses of vehicle control (100% pure sesame oil), melamine (500, 1000, and 2000 mg/kg) or positive control (N-ethyl-N-nitrosourea, ENU, 20 mg/kg) by oral gavage. Peripheral blood was sampled pre-dose (day -1) and at time points up to day 60. Pig-a mutant frequencies were determined in total red blood cells (RBCs) and reticulocytes (RETs) as RBC(CD59-) and RET(CD59-) frequencies, on days -1, 15, 29 and 60, and micronucleus frequencies were measured in RETs on day 4. No significant increases in RBC(CD59-) or RET(CD59-) frequencies were observed for the melamine-treated group at any of the time points studied, but the positive control, ENU, induced statistically significant increases compared with the vehicle control. Similar results were obtained in the micronucleus assay. Melamine did not induce statistically significant increases in %MN-RET. In the bacterial reverse mutation assay, melamine was tested from 62.5 to 1000 μg/plate in tester strains TA97a, TA98, TA100, TA102, and TA1535, with and without metabolic activation, and no evidence of toxicity or mutagenicity was observed at any dose tested. In the in vitro CBMN assay, in Chinese hamster ovary (CHO) cells, melamine was tested (75, 150, and 300 μg/mL) in the presence and absence of S9 mix, and no positive increases in the number of cells containing micronuclei were seen. These results suggest that melamine does not exhibit significant genotoxic potential. These data could be valuable for risk assessment purposes and also for further characterizing the new in vivoPig-a gene mutation assay.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumazid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin -sulfat (Salz), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Melamin, 99%
Sigma-Aldrich
Natriumazid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Methylmethansulfonat, 99%
Sigma-Aldrich
Streptomycin -sulfat (Salz), powder
Sigma-Aldrich
Natriumazid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Natriumazid, BioXtra
Sigma-Aldrich
Anthranilsäure, reagent grade, ≥98%
Sigma-Aldrich
2-Aminoanthracen, 96%
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
N-Nitroso-N-ethylurea, ISOPAC®
Supelco
Melamin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Streptomycin -sulfat (Salz), powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
2-Nitrofluoren, 98%
Supelco
Melamin, analytical standard
Sigma-Aldrich
Anthranilsäure, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
N-Nitroso-N-ethylurea, Bulk package
USP
Melamin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Supelco
Streptomycin -Lösung, ~1 mg/mL in 1 mM EDTA, analytical standard
Supelco
Methylmethansulfonat, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Cyclophosphamid, European Pharmacopoeia (EP) Reference Standard
Streptomycinsulfat, European Pharmacopoeia (EP) Reference Standard