Merck
  • Formulation and characterization of ORMOSIL particles loaded with budesonide for local colonic delivery.
Formulation and characterization of ORMOSIL particles loaded with budesonide for local colonic delivery.
International journal of pharmaceutics (2015-02-25)
Vesna Petrovska-Jovanovska, Nikola Geskovski, Maja Simonoska Crcarevska, Oya Memed, Gjorgji Petruševski, Marina Chachorovska, Marija Petrusevska, Ana Poceva-Panovska, Kristina Mladenovska, Sonja Ugarkovic, Marija Glavas-Dodov
ZUSAMMENFASSUNG
In this study, hybrid silica xerogel particles were developed as carriers of budesonide (BDS) for efficient local treatment of inflammatory bowel diseases (IBD). Organically modified silica particles (ORMOSILs) were prepared by co-condensation of 3-aminopropyltriethoxysilane (APTES) and tetraethyl orthosilicate (TEOS) by an ambient temperature acid catalysed sol-gel process followed by spray-drying. Formulation for preparation of BDS-loaded particles was optimized and their physicochemical parameters and drug release profiles were evaluated in vitro. Optimal formulation had a small particle size (mean diameter of 1.45±0.02μm) with unimodal narrow size distribution and high encapsulation efficiency (98.0 ± 1.85%). Due to the positive surface charge originated from amino group of APTES, ORMOSILs showed excessive mucoadhesiveness in comparison to native TEOS particles. The drug release decreased with increasing pH from 2.0 to 7.4. In order to avoid undesirable erroneous performance in the upper GI tract, particles were additionally coated with Eudragit(®) FS 30D, as a barrier to the drug release at pH range from 2.0 to 7.0. After Eudragit(®) FS 30D coating, the release of BDS in acidic media was sustained, while no significant differences in drug release were observed at pH 7.4. In conclusion, pH-responsive ORMOSILs showed great potential for efficient BDS delivery to the colon region.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Tetraethylorthosilikat, reagent grade, 98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, 99%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Tetraethylorthosilikat, ≥99.0% (GC)
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, ≥98.0%
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, ≥98%
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Tetraethylorthosilikat, 99.999% trace metals basis
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%