Merck
  • Trends and predictors of smoking cessation after percutaneous coronary intervention (from Olmsted County, Minnesota, 1999 to 2010).
Trends and predictors of smoking cessation after percutaneous coronary intervention (from Olmsted County, Minnesota, 1999 to 2010).
The American journal of cardiology (2014-12-30)
Ondrej Sochor, Ryan J Lennon, Juan Pablo Rodriguez-Escudero, John F Bresnahan, Ivana Croghan, Virend K Somers, Francisco Lopez-Jimenez, Quinn Pack, Randal J Thomas
ZUSAMMENFASSUNG
Smoke-free ordinance implementation and advances in smoking cessation (SC) treatment have occurred in the past decade; however, little is known about their impact on SC in patients with coronary artery disease. We conducted a retrospective cohort study of 2,306 consecutive patients from Olmsted County, Minnesota, who underwent their first percutaneous coronary intervention (PCI) from 1999 to 2009, and assessed the trends and predictors of SC after PCI. Smoking status was ascertained by structured telephone survey 6 and 12 months after PCI (ending in 2010). The prevalence of smoking in patients who underwent PCI increased nonsignificantly from 20% in 1999 to 2001 to 24% in 2007 to 2009 (p = 0.14), whereas SC at 6 months after PCI decreased nonsignificantly from 50% (1999 to 2001) to 49% (2007 to 2009), p = 0.82. The 12-month quit rate did not change significantly (48% in 1999 to 2001 vs 56% in 2007 to 2009, p = 0.38), even during the time periods after the enactment of smoke-free policies. The strongest predictor of SC at 6 months after PCI was participation in cardiac rehabilitation (odds ratio [OR] 3.17, 95% confidence interval [CI] 2.05 to 4.91, p <0.001), older age (OR 1.42 per decade, 95% CI 1.16 to 1.73, p <0.001), and concurrent myocardial infarction at the time of PCI (OR 1.77, 95% CI 1.18 to 2.65, p = 0.006). One-year mortality was lower in the group of smokers compared with never smokers (3% vs 7%, p <0.001). In conclusion, SC rates have not improved after PCI over the past decade in our cohort, despite the presence of smoke-free ordinances and improved treatment strategies. Improvements in delivery of systematic services aimed at promoting SC (such as cardiac rehabilitation) should be part of future efforts to improve SC rates after PCI.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.