Merck
  • Challenging core-shell stationary phases with the separation of closely related anti-cancer compounds: performance studies and application to drug quantitation in cell cultures with multi-well plate clean-up.
Challenging core-shell stationary phases with the separation of closely related anti-cancer compounds: performance studies and application to drug quantitation in cell cultures with multi-well plate clean-up.
Journal of chromatography. A (2014-09-06)
Víctor González-Ruiz, Ana I Olives, M Antonia Martín
ZUSAMMENFASSUNG
In the present paper, we compare the chromatographic behaviour of six different stationary phases (5 μm fully porous and 2.6 μm core-shell particles, featuring octadecyl-, pentafluorophenyl-propyl- and phenyl-hexyl- functionalizations) for the separation of camptothecin and a family of closely related analogues of the natural anti-cancer drug luotonin A under different experimental conditions (organic solvent nature and proportion, temperature and presence of modifiers). We found two differentiated behaviours for the phenyl-hexyl-functionalized supports depending on the temperature. The efficiency and kinetic performance of fully porous vs. core-shell particles was compared by means of van Deemter and kinetic plots, using an optimized conventional HPLC system. Columns packing core-shell particles afforded improved efficiencies, permeabilities, analysis times and consumption of solvents. Nevertheless, our optimized system was not able to release the full intrinsic efficiencies of these columns. The replacement of acetonitrile with methanol had a dramatic effect in the behaviour of the pentafluorophenyl-propyl stationary phase. The aggregate of these results led us to select the core-shell C-18 stationary phase along with a H2O:MeCN gradient for the rapid and sustainable quantitation of the anti-tumour agents in cell cultures. A simple and rapid clean-up step was optimized using 96-well deproteinization and delipidation plates. The validation of the proposed analytical method showed excellent figures of merit. The combination of this sample pre-treatment procedure with the separation on core-shell particles yielded a fast methodology able to process a large number of samples with minimal waste generation and environmental impact.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
tert-Butylmethylether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
tert-Butylmethylether, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
tert-Butylmethylether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
tert-Butylmethylether, reagent grade, ≥98%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis