Merck
  • Surfactant protein C metabolism in human infants and adult patients by stable isotope tracer and mass spectrometry.
Surfactant protein C metabolism in human infants and adult patients by stable isotope tracer and mass spectrometry.
Analytical and bioanalytical chemistry (2014-09-04)
Manuela Simonato, Aldo Baritussio, Barbara Pioselli, Carlo Ori, Silvia Catinella, Virgilio P Carnielli, Paola E Cogo
ZUSAMMENFASSUNG
Surfactant protein C (SP-C) is deemed as the surfactant protein most specifically expressed in type II alveolar epithelial cells and plays an important role in surfactant function. SP-C turnover in humans and its meaning in the clinical context have never been approached. In this study, we used mass spectrometry to investigate SP-C turnover in humans. We studied four infants and eight adults requiring mechanical ventilation. All patients had no lung disease. Patients received a 24-h continuous infusion of (13)C-leucine as precursor of SP-C, and serial tracheal aspirates and plasma samples were obtained every 6 h till 48 h. SP-C was isolated from tracheal aspirates by sorbent-phase chromatography. (13)C-leucine SP-C enrichment could be successfully measured in three infant and in four adult samples by using mass spectrometry coupled with a gas chromatographer. Median SP-C fractional synthesis rate, secretion time, and peak time were 15.7 (14.1-27.5)%/day, 6.0 (4.7-11.5) h, and 24 (20-27) h. In conclusion, this study shows that it is feasible to accurately determine SP-C turnover in humans by stable isotopes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
L-Leucin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
L-Leucin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Hexan, anhydrous, 95%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer