Merck
  • Characterization of secreted proteins of 2-cell mouse embryos cultured in vitro to the blastocyst stage with and without protein supplementation.
Characterization of secreted proteins of 2-cell mouse embryos cultured in vitro to the blastocyst stage with and without protein supplementation.
Journal of assisted reproduction and genetics (2014-03-25)
Tanya Burch, Liang Yu, Julius Nyalwidhe, Jose A Horcajadas, Silvina Bocca, R James Swanson, Sergio Oehninger
ZUSAMMENFASSUNG
To identify the secreted proteins of murine embryos grown in vitro. Two-cell mouse embryos (n=432) were randomly allocated to culture to the blastocyst stage in protein-free and in protein-supplemented (3 % BSA) media. Proteins were separated by SDS-PAGE; bands were visualized by coomassie staining, followed by in-gel trypsin digestion and liquid chromatography-tandem mass spectrometry. RT-PCR and confocal microscopy were used to confirm gene/protein expression in blastocysts. Of all individually identified proteins, 34 and 23 were found in embryos cultured without and with BSA, respectively, and 20 were common. Identified proteins having an N-terminal secretory sequence or transmembrane domains located on the extracellular backbone were postulated as secreted proteins. Gene and protein expression for two selected molecules were confirmed. Functional analysis revealed over-represented processes related to lipid metabolism, cyclase activity, and cell adhesion/membrane functions. This study provided evidence to further characterize secreted proteins by mouse embryos grown from the 2-cell to the blastocyst stage in vitro. Because of homology between murine and human, these results may provide information to be translated to the clinical setting.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Jodacetamid, BioUltra
Sigma-Aldrich
Jodacetamid, Single use vial of 56 mg
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Jodacetamid, ≥99% (NMR), crystalline
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidiumbromid, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Supelco
Trifluoressigsäure, analytical standard
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethidiumbromid, ~95% (HPLC)
SAFC
Jodacetamid
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Ethidiumbromid -Lösung, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Ameisensäure -Lösung, BioUltra, 1.0 M in H2O