Merck
  • Effects of dopaminergic agents on progression of naturally occurring myopia in albino guinea pigs (Cavia porcellus).
Effects of dopaminergic agents on progression of naturally occurring myopia in albino guinea pigs (Cavia porcellus).
Investigative ophthalmology & visual science (2014-10-02)
Liqin Jiang, Keli Long, Frank Schaeffel, Xiangtian Zhou, Yibo Zheng, Huangfang Ying, Fan Lu, William K Stell, Jia Qu
ZUSAMMENFASSUNG
Disruption of dopaminergic signaling has been implicated in the abnormalities of ocular development in albinism, and many experiments have shown that retinal dopamine is a major regulator of postnatal eye growth and myopia in animal models. Therefore, in the present study we investigated whether progressive myopia, which can occur in albino guinea pigs without experimental manipulation of visual conditions, is affected by dopaminergic agents. Two-week-old albino guinea pigs, selected for being myopic (range refractive error [RE], -2 to -10 diopters [D]), received unilateral peribulbar injections of apomorphine (nonselective dopamine receptor agonist; 0, 7.5, 25, 75, 250, 750, and 2500 ng; n = 112), SKF38393 (D1-like agonist; 0, 10, 100, 1000 ng; n = 63), SCH23390 (D1-like antagonist; 0, 2500 ng; n = 27), quinpirole (D2-like agonist; 0, 10, 100, 1000 ng; n = 58), or sulpiride (D2-like antagonist; 0, 2500 ng; n = 24) once a day for four weeks. One noninjected group (n = 19) served as untreated control. Refractive states and axial dimensions of the eyes were measured without cycloplegia or general anesthetic, using eccentric infrared photoretinoscopy and A-scan ultrasonography, respectively, before treatment, and after 2 and 4 weeks of treatment. The main drug effects were analyzed by paired t-test or 2-way repeated measures ANOVA, as required. The naturally occurring progression of myopic RE was inhibited by apomorphine at relatively high doses (250 and 750 ng), SKF38393 at 100 ng (D1-like agonist), and sulpiride at 2500 ng (D2-like antagonist), but promoted by apomorphine at a lower dose (25 ng), quinpirole at 100 ng (D2-like agonist), and SCH23390 at 2500 ng (D1-like antagonist). All drugs affected primarily vitreous chamber depth, rather than anterior segment dimensions. Our data suggest that the activation of D1-like receptors inhibits, whereas activation of D2-like receptors promotes, progressive myopia in this animal model. The robust effects of antagonists suggest that ocular dopamine receptors in these albinos may be in a chronic state of partial excitation. The precise location and identity of the receptors responsible for these effects remain to be determined.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
USP
Ascorbinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Supelco
L-Ascorbinsäure, analytical standard
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbinsäure, FCC, FG
Sigma-Aldrich
L-Ascorbinsäure, BioUltra, ≥99.5% (RT)
Ascorbinsäure, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethylsulfoxid, analytical standard
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
(±)-Sulpirid
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)