Merck
EGF up-regulates miR-31 through the C/EBPβ signal cascade in oral carcinoma.
PloS one (2014-09-18)
Wen-Cheng Lu, Shou-Yen Kao, Cheng-Chieh Yang, Hsi-Feng Tu, Cheng-Hsien Wu, Kuo-Wei Chang, Shu-Chun Lin
ZUSAMMENFASSUNG
Oral squamous cell carcinoma (OSCC) is one of the most prevalent carcinomas worldwide. MicroRNAs (miRNAs) are short, non-coding RNAs that regulate gene expression and modulate physiological or pathological processes including OSCC carcinogenesis. miR-31 has been found to be up-regulated in OSCC and to act as an oncogenic miRNA. However, the molecular mechanism underlying miR-31 up-regulation in OSCC is still obscure. The activation of epidermal growth factor receptor (EGFR) signaling axis plays key roles in driving oral carcinogenesis. Our screening identified that there is up-regulation of miR-31, miR-181b and miR-222 in OSCC cells following EGF treatment. Subsequent analysis showed that EGF treatment led to AKT activation, which then resulted in miR-31 up-regulation. Moreover, EGF treatment and the AKT activation induced by exogenous expression up-regulated C/EBPβ expression. The miR-31 up-regulation induced by EGF was abrogated by AKT inhibition or by the knockdown of C/EBPβ expression. In OSCC cell subclones stably overexpressing the functional isoform of C/EBPβ, miR-31 expression was up-regulated. Curcumin is a natural ingredient exhibiting anti-cancer potential. It was found that curcumin attenuated AKT activation and the up-regulation of C/EBPβ and miR-31 caused by EGF stimulation in OSCC cells. Lastly, concordance across the expression of EGFR, the expression of C/EBPβ and the expression of miR-31 in OSCC tissues was found. This study describes a novel scenario where the up-regulation of miR-31 expression in OSCC is, at least in part, a consequence of EGFR oncogenic activation. Although the AKT activation and C/EBPβ expression after EGF treatment might not be directly linked, both events are the crucial mediators underlying miR-31 up-regulation in the EGFR signaling axis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Curcumin, analytical standard
USP
Curcumin, United States Pharmacopeia (USP) Reference Standard
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder
Supelco
Curcumin, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC)
Curcumin, primary reference standard
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®