Merck
Juvenile obesity enhances emotional memory and amygdala plasticity through glucocorticoids.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2015-03-06)
Chloé Boitard, Mouna Maroun, Frédéric Tantot, Amandine Cavaroc, Julie Sauvant, Alain Marchand, Sophie Layé, Lucile Capuron, Muriel Darnaudery, Nathalie Castanon, Etienne Coutureau, Rose-Marie Vouimba, Guillaume Ferreira
ZUSAMMENFASSUNG
In addition to metabolic and cardiovascular disorders, obesity is associated with adverse cognitive and emotional outcomes. Its growing prevalence during adolescence is particularly alarming since recent evidence indicates that obesity can affect hippocampal function during this developmental period. Adolescence is a decisive period for maturation of the amygdala and the hypothalamic-pituitary-adrenal (HPA) stress axis, both required for lifelong cognitive and emotional processing. However, little data are available on the impact of obesity during adolescence on amygdala function. Herein, we therefore evaluate in rats whether juvenile high-fat diet (HFD)-induced obesity alters amygdala-dependent emotional memory and whether it depends on HPA axis deregulation. Exposure to HFD from weaning to adulthood, i.e., covering adolescence, enhances long-term emotional memories as assessed by odor-malaise and tone-shock associations. Juvenile HFD also enhances emotion-induced neuronal activation of the basolateral complex of the amygdala (BLA), which correlates with protracted plasma corticosterone release. HFD exposure restricted to adulthood does not modify all these parameters, indicating adolescence is a vulnerable period to the effects of HFD-induced obesity. Finally, exaggerated emotional memory and BLA synaptic plasticity after juvenile HFD are alleviated by a glucocorticoid receptor antagonist. Altogether, our results demonstrate that juvenile HFD alters HPA axis reactivity leading to an enhancement of amygdala-dependent synaptic and memory processes. Adolescence represents a period of increased susceptibility to the effects of diet-induced obesity on amygdala function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lithiumchlorid, ACS reagent, ≥99%
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Lithiumchlorid, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Lithiumchlorid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Lithiumchlorid, ReagentPlus®, 99%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Benzaldehyd, ReagentPlus®, ≥99%
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Lithiumchlorid -Lösung, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Benzaldehyd, purified by redistillation, ≥99.5%
Sigma-Aldrich
Lithiumchlorid, Molecular Biology, ≥99%
Sigma-Aldrich
Lithiumchlorid, powder, ≥99.98% trace metals basis
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Mifepriston, ≥98%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Elektrolyt -Lösung, nonaqueous, 2 M LiCl in ethanol
Supelco
Benzaldehyd, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Isoamylacetat, natural, ≥97%, FCC, FG
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%