Merck
  • Diclofenac toxicity in human intestine ex vivo is not related to the formation of intestinal metabolites.
Diclofenac toxicity in human intestine ex vivo is not related to the formation of intestinal metabolites.
Archives of toxicology (2014-04-29)
Xiaoyu Niu, Inge A M de Graaf, Miriam Langelaar-Makkinje, Peter Horvatovich, Geny M M Groothuis
ZUSAMMENFASSUNG
The use of diclofenac (DCF), a nonsteroidal anti-inflammatory drug, is associated with a high prevalence of gastrointestinal side effects. In vivo studies in rodents suggested that reactive metabolites of DCF produced by the liver or the intestine might be responsible for this toxicity. In the present study, precision-cut intestinal slices (PCIS) prepared from the jejunum of 18 human donors were used as an ex vivo model to investigate whether DCF intestinal metabolites are responsible for its intestinal toxicity in man. PCIS were incubated with a concentration range of DCF (0-600 µM) up to 24 h. DCF (≥400 µM) caused direct toxicity to the intestine as demonstrated by ATP depletion, morphological damage, caspase 3 activation, and lactate dehydrogenase leakage. Three main metabolites produced by PCIS (4'-hydroxy DCF, 5-hydroxy DCF, and DCF acyl glucuronide) were detected by HPLC. Protein adducts were detected by immunohistochemical staining and showed correlation with the intestinal metabolites. DCF induced similar toxicity to each of the samples regardless of the variation in metabolism among them. Less metabolites were produced by slices incubated with 400 µM DCF than with 100 µM DCF. The addition of the metabolic inhibitors such as ketoconazole, cimetidine, or borneol decreased the metabolite formation but increased the toxicity. The results suggest that DCF can induce intestinal toxicity in human PCIS directly at therapeutically relevant concentrations, independent of the reactive metabolites 4'-OH DCF, 5-OH DCF, or diclofenac acylglucuronide produced by the liver or formed in the intestine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Supelco
Ammoniumformat, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammoniumformat, reagent grade, 97%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis