Merck
Nanocarriers and the delivered drug: effect interference due to intravenous administration.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2014-06-26)
Maria A Vlasova, Jussi Rytkönen, Joakim Riikonen, Olga S Tarasova, Juha Mönkäre, Miia Kovalainen, Ale Närvänen, Jarno Salonen, Karl-Heinz Herzig, Vesa-Pekka Lehto, Kristiina Järvinen
ZUSAMMENFASSUNG
Intravenously administered nanocarriers are widely studied to improve the delivery of various therapeutic agents. However, recent in vivo studies have demonstrated that intravenously administered nanocarriers that do not contain any drug may affect cardiovascular function. Here we provide an example where the drug and the nanocarrier both affect the same cardiovascular parameters following intravenous administration. The peptide ghrelin antagonist (GhA) increases arterial pressure, while thermally hydrocarbonized porous silicon nanoparticles (THCPSi) transiently decrease it, as assessed with radiotelemetry in conscious rats. As a result, intravenous administration of GhA-loaded THCPSi nanoparticles partially antagonized GhA activity: arterial pressure was not increased. When the cardiovascular effects of GhA were blocked with atenolol pretreatment, GhA-loaded nanoparticles reduced arterial pressure to similar extent as drug-free nanoparticles. These data indicate that the biological activity of a drug delivered within a nanocarrier may be obscured by the biological responses induced by the nanocarrier itself.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Kaliumiodid, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Fluorwasserstoffsäure, ACS reagent, 48%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Kaliumiodid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Natriummetabisulfit, ReagentPlus®, ≥99%
Sigma-Aldrich
Kaliumiodid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
Fluorwasserstoffsäure, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Natriummetabisulfit, reagent grade, 97%
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Natriummetabisulfit, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Kaliumiodid, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Kaliumiodid, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Supelco
Trifluoressigsäure, analytical standard
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Supelco
Natriummetabisulfit, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Kaliumiodid, ≥99.99% trace metals basis