Merck
  • Hepatic microsomal thiol methyltransferase is involved in stereoselective methylation of pharmacologically active metabolite of prasugrel.
Hepatic microsomal thiol methyltransferase is involved in stereoselective methylation of pharmacologically active metabolite of prasugrel.
Drug metabolism and disposition: the biological fate of chemicals (2014-04-16)
Miho Kazui, Katsunobu Hagihara, Takashi Izumi, Toshihiko Ikeda, Atsushi Kurihara
ZUSAMMENFASSUNG
Prasugrel, a thienopyridine antiplatelet drug, is converted in animals and humans to the pharmacologically active metabolite R-138727 [(2Z)-{1-[(1RS)-2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-4-sulfanylpiperidin-3-ylidene}ethanoic acid], which has two chiral centers, occurring as a mixture of four isomers. The RS and RR isomers are more active than the SS and SR isomers (RS > RR > > SR = SS). The pharmacologically active metabolite is further metabolized to an S-methylated metabolite that is the major identified inactive metabolite in humans. In rat, dog, and human liver microsomes supplemented with S-adenosyl methione, the SS and SR isomers of the active metabolite were extensively S-methylated while the RS and RR isomers were not. Addition of 2,3-dichloromethyl benzylamine (50 µM) completely inhibited the S-methylation reaction, indicating that the microsomal and cytosolic thiol methyltransferase but not the cytosolic thiopurine S-methyltransferase is involved in the methylation. The hepatic intrinsic clearance values for methylation of the RS, RR, SS, and SR isomers (ml/min/kg) were 0, 0, 40.4, and 37.6, respectively, in rat liver microsomes, 0, 0, 11.6, and 2.5, respectively, in dog liver microsomes, and 0, 0, 17.3, and 17.7, respectively, in human liver microsomes, indicating that the RS and RR isomers are not methylated in vitro and that the methylation of SS and SR isomers is high with rat > human > dog. This finding in vitro agreed well with the in vivo observation in rats and dogs, where the S-methylated SS and SR isomers were the major metabolites in the plasma whereas negligible amounts of S-methylated RS and RR isomers were detected after intravenous administration of the pharmacologically active metabolites.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, suitable for HPLC
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Wasser, Deionized
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Supelco
Wasser, suitable for ion chromatography
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Supelco
Wasser, for TOC analysis
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotidphosphat Hydrat
Sigma-Aldrich
Wasser, BioPerformance Certified
Supelco
Glutathion, Pharmaceutical Secondary Standard; Certified Reference Material
Wasser-Dichtestandard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Wasser, ACS reagent
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
Supelco
Wasser, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Deuterium, 99.8 atom % D
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Wasser, PCR Reagent
Sigma-Aldrich
Wasser, tested according to Ph. Eur.
Sigma-Aldrich
3-Methoxybenzoesäure, ReagentPlus®, 99%
Supelco
Wasser, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotidphosphat Natriumsalz, pkg of 5 mg (per vial)
Glutathion, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Deuterium, 99.9 atom % D
Sigma-Aldrich
Wasser, endotoxin, free
Wasser-Dichtestandard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
ω-Brom-3-methoxyacetophenon, 98%
Supelco
Dichte Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality