Merck
ArsH from Synechocystis sp. PCC 6803 reduces chromate and ferric iron.
FEMS microbiology letters (2014-05-28)
Xi-Mei Xue, Yu Yan, Hui-Juan Xu, Ning Wang, Xiao Zhang, Jun Ye
ZUSAMMENFASSUNG
ArsH is widely distributed in bacteria, and its function remains to be characterized. In this study, we investigated the function of ArsH from Synechocystis sp. PCC 6803. The inactivation of arsH by insertion of a kanamycin-resistance gene in Synechocystis sp. PCC 6803 resulted in the decrease of arsenic and chromium accumulation compared with the wild type. ArsH expression in Escherichia coli strain Rosetta increased its resistance to chromate by reducing chromate in the medium and cells to chromium (III). In addition, ArsH in Rosetta conferred resistance to arsenic. The purified Synechocystis ArsH was able to reduce chromate and ferric iron at the expense of NADPH. Nonlinear regression values of K0.5 for chromate and ferric iron were 71.9 ± 17.8 μM and 59.3 ± 13.8 μM, respectively. The expression level of arsH was induced by arsenite and arsenate, but not chromate or ferric iron. Our results suggest that Synechocystis ArsH had no substrate specificities and shared some biochemical properties that other enzymes possessed. ArsH may be involved in coordinating oxidative stress response generated by arsenic.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salpetersäure, ACS reagent, 70%
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Salpetersäure, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
1,10-Phenanthrolin, ≥99%
Sigma-Aldrich
Salpetersäure, red, fuming, HNO3 >90 %
Sigma-Aldrich
1,5-Diphenylcarbazid, ACS reagent
Sigma-Aldrich
Nitrilotriessigsäure, Sigma Grade, ≥99%
Supelco
Salpetersäure-Konzentrat, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Isopropyl β-D-1-Thiogalactopyranosid, ≥99% (TLC)
Sigma-Aldrich
Salpetersäure, ACS reagent, ≥90.0%
Sigma-Aldrich
Isopropyl-β-D-thiogalactopyranosid -Lösung, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
Chrom, powder, ≥99% trace metals basis, <45 μm
Supelco
Nitrilotriessigsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nitrilotriessigsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
1,5-Diphenylcarbazid, reagent grade
Supelco
1,5-Diphenylcarbazid, reag. Ph. Eur., ≥98.0%
Sigma-Aldrich
Chrom, powder, 99.5%, −100 mesh
SAFC
Isopropyl β-D-1-Thiogalactopyranosid
Sigma-Aldrich
Chrom, chips, 99.995% trace metals basis
Sigma-Aldrich
Chrom, chips, thickness ~2 mm, 99.5%
Chrom, rod, 25mm, diameter 2.0mm, 99.7+%
Sigma-Aldrich
Nitrilotriessigsäure, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Salpeter-14N-Säurelösung, ~10 N in H2O, 99.99 atom % 14N
Chrom, foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Chrom, rod, 50mm, diameter 2.0mm, 99.7+%
Chrom, rod, 10mm, diameter 2.0mm, 99.7+%
Chrom, bar, 100mm x 2mm x 2mm, 99.7+%
Chrom, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Chrom, microfoil, disks, 10mm, thinness 0.025μm, specific density 18.21μg/cm2, permanent mylar 3.5μm support, 99.99+%