Merck
Human amniotic fluid stem cells: neural differentiation in vitro and in vivo.
Cell and tissue research (2014-05-03)
Tullia Maraldi, Laura Bertoni, Massimo Riccio, Manuela Zavatti, Gianluca Carnevale, Elisa Resca, Marianna Guida, Francesca Beretti, Giovanni B La Sala, Anto De Pol
ZUSAMMENFASSUNG
The successful integration of stem cells after their implantation into the brain has become a central issue in modern neuroscience. In this study, we test the neural differentiation potential of c-Kit(+)/Oct-4(+) human amniotic fluid stem cells (hAFSCs) in vitro and their survival and integration in vivo. hAFSCs were induced towards neural differentiation and specific markers (GFAP, β-III tubulin, CNPase, MAP2, NeuN, synapsines, S100, PMP22) were detected by immunofluorescence and Western blot analysis. Glial proteins were expressed as early as 2 weeks after the initial differentiation stimulus, whereas neuronal markers started to appear from the third week of differentiation under culturing conditions of high cell density. This timeline suggested that glial cells possessed a promoting role in the differentiation of hAFSCs towards a neuronal fate. hAFSCs were then implanted into the lateral ventricle of the brain of 1-day-old rats, since neuronal development occurs up to 1 month after birth in this animal model. Our data showed that hAFSCs survived for up to 6 weeks post-implantation, were integrated into various areas of the central nervous system and migrated away from the graft giving rise to mature neurons and oligodendrocytes. We conclude that hAFSCs are able to differentiate and integrate into nervous tissue during development in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
SAFC
L-Glutamin
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)