Merck
  • Carbon-based nanomaterials accelerate arteriolar thrombus formation in the murine microcirculation independently of their shape.
Carbon-based nanomaterials accelerate arteriolar thrombus formation in the murine microcirculation independently of their shape.
Journal of applied toxicology : JAT (2014-02-18)
Martin Holzer, Peter Bihari, Marc Praetner, Bernd Uhl, Christoph Reichel, Janos Fent, Minnamari Vippola, Susan Lakatos, Fritz Krombach
ZUSAMMENFASSUNG
Although carbon-based nanomaterials (CBNs) have been shown to exert prothrombotic effects in microvessels, it is poorly understood whether CBNs also have the potential to interfere with the process of leukocyte-endothelial cell interactions and whether the shape of CBNs plays a role in these processes. Thus, the aim of this study was to compare the acute effects of two differently shaped CBNs, fiber-shaped single-walled carbon nanotubes (SWCNT) and spherical ultrafine carbon black (CB), on thrombus formation as well as on leukocyte-endothelial cell interactions and leukocyte transmigration in the murine microcirculation upon systemic administration in vivo. Systemic administration of both SWCNT and CB accelerated arteriolar thrombus formation at a dose of 1 mg kg(-1) body weight, whereas SWCNT exerted a prothrombotic effect also at a lower dose (0.1 mg kg(-1) body weight). In vitro, both CBNs induced P-selectin expression on human platelets and formation of platelet-granulocyte complexes. In contrast, injection of fiber-shaped SWCNT or of spherical CB did not induce leukocyte-endothelial cell interactions or leukocyte transmigration. In vitro, both CBNs slightly increased the expression of activation markers on human monocytes and granulocytes. These findings suggest that systemic administration of CBNs accelerates arteriolar thrombus formation independently of the CBNs' shape, but does not induce leukocyte-endothelial cell interactions or leukocyte transmigration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid, tablet