Merck
Inhibition screening method of microsomal UGTs using the cocktail approach.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2015-02-17)
Julieta Gradinaru, Stéphanie Romand, Laurent Geiser, Pierre-Alain Carrupt, Dany Spaggiari, Serge Rudaz
ZUSAMMENFASSUNG
A rapid method for the simultaneous determination of the in vitro activity of the 10 major human liver UDP-glucuronosyltransferase (UGT) enzymes was developed based on the cocktail approach. Specific substrates were first selected for each UGT: etoposide for UGT1A1, chenodeoxycholic acid for UGT1A3, trifluoperazine for UGT1A4, serotonin for UGT 1A6, isoferulic acid for UGT1A9, codeine for UGT2B4, azidothymidine for UGT2B7, levomedetomidine for UGT2B10, 4-hydroxy-3-methoxymethamphetamine for UGT2B15 and testosterone for UGT2B17. Optimal incubation conditions, including time-based experiments on cocktail metabolism in pooled HLMs that had been performed, were then investigated. A 45-min incubation period was found to be a favorable compromise for all the substrates in the cocktail. Ultra-high pressure liquid chromatography coupled to an electrospray ionization time-of-flight mass spectrometer was used to separate the 10 substrates and their UGT-specific glucuronides in less than 6 min. The ability of the cocktail to highlight the UGT inhibitory potential of xenobiotics was initially proven by using well-known UGT inhibitors (selective and nonselective) and then by relating some of the screening results obtained by using the cocktail approach with morphine glucuronidation (substrate highly glucuronidated by UGT 2B7). All the results were in agreement with both the literature and with the expected effect on morphine glucuronidation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Perchlorsäure, ACS reagent, 70%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis