Merck
  • Novel pyridinium-based tags: synthesis and characterization for highly efficient analysis of thiol-containing peptides by mass spectrometry.
Novel pyridinium-based tags: synthesis and characterization for highly efficient analysis of thiol-containing peptides by mass spectrometry.
The Analyst (2014-11-26)
Xiaoqiang Qiao, Yanjun Yang, Shijia Liu, Shiheng Chen, Xin Wang, Guangyue Li, Hongyuan Yan, Xun Yang
ZUSAMMENFASSUNG
In this study, a novel type of pyridinium-based tags, 1-[3-[(2-iodo-1-oxoethyl)amino]propyl]-4-methylpyridinium bromide (IMP) and 1-[3-[(2-iodo-1-oxoethyl)amino]propyl]-4-propylpyridinium bromide (IPP), were designed, synthesized, and applied to the derivatization of thiol-containing peptides. With model peptides as the sample, the labeling efficiency and the stability of the peptide derivatives were investigated. The results indicate that nearly 100% derivatization yield was achieved with the developed tags and the peptide derivatives were stable at room temperature for at least one week. Furthermore, improved ionization efficiency and increased charge states were achieved via both matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MS) and electrospray ionization (ESI) MS, of which IPP exhibited the more obvious improvement of ionization efficiency. Further analysis of tryptic digests of bovine serum albumin (BSA) and α-transferrin, showed that increased identification efficiency of the thiol-containing peptides was achieved by combination with IMP or IPP derivatization. For example, the identification efficiency of the thiol-containing peptides of α-transferrin increased more than 42% upon combination with the IMP or IPP derivatives. We anticipate the novel tags are promising for highly efficient thiol-containing peptide identification in proteome research, especially for low concentrations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Tris(2-carboxyethyl)phosphin -hydrochlorid, powder
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Jodacetamid, BioUltra
Supelco
Tris(2-carboxyethyl)phosphin -hydrochlorid -Lösung, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)
Sigma-Aldrich
Jodacetamid, Single use vial of 56 mg
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Jodacetamid, ≥99% (NMR), crystalline
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
3-Indolessigsäure, 98%
Sigma-Aldrich
Tris(2-carboxyethyl)phosphin -hydrochlorid, BioUltra, ≥98% (NMR)
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
3-Indolessigsäure, suitable for plant cell culture, crystalline
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Supelco
Trifluoressigsäure, analytical standard
Sigma-Aldrich
Iodessigsäure, ≥98.0% (T)
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
Indol-3-essigsäure Natriumsalz, suitable for plant cell culture, BioReagent, ≥98%