Merck
  • Aneuploidy induces profound changes in gene expression, proliferation and tumorigenicity of human pluripotent stem cells.
Aneuploidy induces profound changes in gene expression, proliferation and tumorigenicity of human pluripotent stem cells.
Nature communications (2014-09-10)
Uri Ben-David, Gal Arad, Uri Weissbein, Berhan Mandefro, Adva Maimon, Tamar Golan-Lev, Kavita Narwani, Amander T Clark, Peter W Andrews, Nissim Benvenisty, Juan Carlos Biancotti
ZUSAMMENFASSUNG
Human pluripotent stem cells (hPSCs) tend to acquire genomic aberrations in culture, the most common of which is trisomy of chromosome 12. Here we dissect the cellular and molecular implications of this trisomy in hPSCs. Global gene expression analyses reveal that trisomy 12 profoundly affects the gene expression profile of hPSCs, inducing a transcriptional programme similar to that of germ cell tumours. Comparison of proliferation, differentiation and apoptosis between diploid and aneuploid hPSCs shows that trisomy 12 significantly increases the proliferation rate of hPSCs, mainly as a consequence of increased replication. Furthermore, trisomy 12 increases the tumorigenicity of hPSCs in vivo, inducing transcriptionally distinct teratomas from which pluripotent cells can be recovered. Last, a chemical screen of 89 anticancer drugs discovers that trisomy 12 raises the sensitivity of hPSCs to several replication inhibitors. Together, these findings demonstrate the extensive effect of trisomy 12 and highlight its perils for successful hPSC applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Methylenblau, certified by the BSC
Sigma-Aldrich
Methylenblau -Lösung, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Gemcitabin-Hydrochlorid -hydrochlorid, ≥98% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Methylenblau -Lösung, 0.05 wt. % in H2O
Sigma-Aldrich
Methylenblau -Lösung, suitable for microscopy
Sigma-Aldrich
Methylenblau -Lösung, suitable for microbiology
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
USP
Gemcitabin-Hydrochlorid -hydrochlorid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methylenblaulösung 1,4 % (w/v) 95 % Ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Gemcitabin-Hydrochlorid -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
17-(Allylamino)-17-demethoxygeldanamycin, ≥98% (HPLC), solid
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Amsacrine hydrochloride, ≥98% (TLC), powder
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)