Merck
  • Stir fabric phase sorptive extraction for the determination of triazine herbicides in environmental waters by liquid chromatography.
Stir fabric phase sorptive extraction for the determination of triazine herbicides in environmental waters by liquid chromatography.
Journal of chromatography. A (2014-12-24)
Mercedes Roldán-Pijuán, Rafael Lucena, Soledad Cárdenas, Miguel Valcárcel, Abuzar Kabir, Kenneth G Furton
ZUSAMMENFASSUNG
Stir fabric phase sorptive extraction (SFPSE), which integrates sol-gel hybrid organic-inorganic coated fabric phase sorptive extraction media with a magnetic stirring mechanism, is presented for the first time. Two flexible fabric substrates, cellulose and polyester were used as the host matrix for three different sorbents e.g., sol-gel poly(tetrahydrofuran), sol-gel poly(ethylene glycol), and sol-gel poly(dimethyldiphenylsiloxane). The new microextraction device has been analytically evaluated using triazine herbicides as model compounds. The factors affecting the extraction efficiency of SFPSE have been investigated and the optimal extraction conditions have been determined. Under these optimum conditions, the limits of quantification (LOQs) for sol-gel poly(ethylene glycol) coated SFPSE device in combination with UPLC-DAD for the analysis of the seven triazine herbicides were in the range of 0. 26-1.50μg/L with precision (relative standard deviation) at 2μg/L concentration ranging from 1.4-4.8% (intra-day, n=5) and 6.8-11.8% (inter-day, n=3). Enrichment factors were found between 444 and 1411 (compared to 2000 theoretical maximum). Absolute extraction recoveries were in the range of 22.2-70.5%. The developed method was applied for the determination of selected triazine herbicides from three river water samples. Relative recoveries of the target analytes, in the range from 75 to 126%, were found to be satisfactory. The combination of SFPSE with LC-MS/MS allows the improvement of the method sensitivity to the range from 0.015μg/L to 0.026μg/L with precision better than 10.8% expressed as relative standard deviation (RSD).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)