Merck
Rapid aptasensor capable of simply diagnosing prostate cancer.
Biosensors & bioelectronics (2014-06-29)
Timothy Cha, Sandy Cho, Young Teck Kim, Ji Hoon Lee
ZUSAMMENFASSUNG
Using guanine (G)-rich DNA aptamer-conjugated 6-carboxyfluorescein (6-FAM) capable of rapidly capturing prostate specific antigen (PSA) in human serum, cost-effective and simple biosensor with guanine chemiluminescence detection was developed for early diagnosis of prostate cancer. Free G-rich DNA aptamer-conjugated 6-FAM emits bright light in guanine chemiluminescence reaction based on the principle of chemiluminescent resonance energy transfer (CRET). However, G-rich DNA aptamer-conjugated 6-FAM bound with PSA cannot emit light because PSA acts as a strong interference in CRET between 6-FAM and high-energy intermediate formed from the reaction of 3,4,5-trimethoxylphenylglyoxal (TMPG) and guanine of G-rich DNA aptamer. A chemiluminescent biosensor, developed using the different properties of G-rich DNA aptamer-conjugated 6-FAM in the absence and presence of PSA in guanine chemiluminescence reaction, was able to quantify trace levels of PSA in human serum within 30 min without time-consuming and complicated procedures (e.g., multiple incubation and washings) required for conventional immunoassays operated with expensive and intractable antibodies. The limit of detection of chemiluminescent biosensor having a wide linear dynamic range (1.9-125 ng/ml) was 1.0 ng/ml. The excellent correlation (R=0.985) between chemiluminescent biosensor and conventional enzyme immunoassay indicates that the accurate, precise, and rapid chemiluminescent biosensor can be applied as a new method for early diagnosis of prostate cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
N,N-Dimethylformamid, suitable for HPLC, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
N,N-Dimethylformamid, biotech. grade, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Dimethylformamid, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Tetrapropylammoniumhydroxid -Lösung, 1.0 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
N,N-Dimethylformamid, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.