Merck
  • Application of bar adsorptive microextraction (BAμE) for anti-doping control screening of anabolic steroids in urine matrices.
Application of bar adsorptive microextraction (BAμE) for anti-doping control screening of anabolic steroids in urine matrices.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-08-27)
S M Ahmad, C Almeida, N R Neng, J M F Nogueira
ZUSAMMENFASSUNG
This study proposes a new analytical methodology for the determination of trace levels of testosterone (T) and epitestosterone (E) in urine matrices using bar adsorptive microextraction combined with liquid desorption followed by high-performance liquid chromatography with diode array detection (BAμE-LD/HPLC-DAD). The comparison of different sorbent coatings (five activated carbons, one styrene-divinylbenzene, two modified pyrrolidone, one ciano and one n-vinylpyrrolidone polymers) through BAμE showed that the latter phase presented much higher selectivity and capacity offering multiple mechanisms of interaction. Assays using this phase were performed on 25mL of water samples spiked at the 8.0μg/L level, yielded average recoveries of 92.1 and 93.4% for T and E, respectively, under optimized experimental conditions; BAμE (n-vinylpyrrolidone): 16h (1000rpm), pH 5.5; LD: acetonitrile, 30min under sonication treatment. From the developed analytical methodology, suitable detection limits were achieved (0.4μg/L) and good linear dynamic ranges (1.4-16.0μg/L) with remarkable determination coefficients (r(2)>0.9978). By using the standard addition methodology, the application of the present analytical approach on urine samples revealed good sensitivity. The proposed method, which operated under the floating sampling technology, proved to be a suitable sorption-based static microextraction alternative for screening T, E and the T/E ratio in urine samples for doping control purposes. The methodology showed to be easy to implement, demonstrating good reproducibility, sensitivity and robustness, allowing the possibility to choose the most selective sorbent coating according to the compounds of interest.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Styrol, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%