Merck
  • Liquid chromatographic-tandem mass spectrometric method for the simultaneous determination of alkylphenols polyethoxylates, alkylphenoxy carboxylates and alkylphenols in wastewater and surface-water.
Liquid chromatographic-tandem mass spectrometric method for the simultaneous determination of alkylphenols polyethoxylates, alkylphenoxy carboxylates and alkylphenols in wastewater and surface-water.
Journal of chromatography. A (2014-08-31)
L Ciofi, C Ancillotti, U Chiuminatto, D Fibbi, L Checchini, S Orlandini, M Del Bubba
ZUSAMMENFASSUNG
Four different pellicular stationary phases (i.e. octadecylsilane, octasilane, Phenyl-Hexyl and pentafluorophenyl) were investigated for the chromatographic resolution of alkylphenols (APs), alkylphenols polyethoxylates (APnEOs) and alkylphenoxy carboxylates (APECs) using mixtures of water and organic solvents (i.e. methanol, acetonitrile and tetrahydrofuran) as eluents, in order to obtain their determination by a single LC-MS/MS run. In fact, alkylphenols and alkylphenoxy carboxylates must be analysed in negative ion mode, whereas alkylphenols polyethoxylates undergo ionisation only in positive ion mode, and therefore, two distinct LC-MS/MS analysis are commonly adopted. The best resolution among the aforementioned target analytes was achieved on the pentafluorophenyl column, eluting with an acidified water-acetonitrile-tetrahydrofuran mixture and using the post column addition of an ammonia solution in methanol for the detection of positively ionisable compounds. Under these optimized chromatographic conditions the investigated compounds were determined via a single chromatographic run, with only one polarity switch, in 15min, achieving the following instrumental detection limits: 600pg for AP1EOs, 0.8-14pg for AP2EOs, 10.4-150pg for APs and 4.4-4.8pg for APECs. The chromatographic method was coupled with solid-phase extraction and clean-up procedures and successfully applied to the analysis of wastewater and surface water samples, highlighting mean concentration ranging from 6ng/L for 4-t-OP1EC to 1434ng/L for 4-NP1121EC, depending on the sample analysed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Supelco
Ammoniumformat, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammoniak -Lösung, 7 N in methanol
Sigma-Aldrich
Ammoniumformat, reagent grade, 97%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ammoniumformat, ≥99.995% trace metals basis
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Ammoniumformiat -Lösung, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ammoniak -Lösung, 2.0 M in ethanol
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard