Merck
  • Carotenoids of aleurone, germ, and endosperm fractions of barley, corn and wheat differentially inhibit oxidative stress.
Carotenoids of aleurone, germ, and endosperm fractions of barley, corn and wheat differentially inhibit oxidative stress.
Journal of agricultural and food chemistry (2015-02-24)
Kabo Masisi, William L Diehl-Jones, Joseph Gordon, Donald Chapman, Mohammed H Moghadasian, Trust Beta
ZUSAMMENFASSUNG
The antioxidant potential of carotenoids from aleurone, germ, and endosperm fractions of barley, corn, and wheat has been evaluated. HPLC analysis confirmed the presence of lutein and zeaxanthin carotenoids (nd-15139 μg/kg) in extracts of cereal grain fractions. The antioxidant properties using 2,2-diphenyl-1-picrylhydrazyl, oxygen radical absorbance capacity, 2,2'-azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) assays revealed significantly higher (P<0.001) antioxidant activity in the germ than in the aleurone and endosperm fractions. Using 3-(4,5-dimethylthiazolyl-2)-2,5-diphenyltetrazolium bromide (MTT) assay, 2,2'azobis (2-amidinopropane)dihydrochloride (AAPH)-induced cell loss was effectively reduced by preincubating Caco-2, HT-29, and FHs 74 Int cells with carotenoid extracts. Moreover, carotenoid extracts reduced (P<0.001) AAPH-induced intracellular oxidation in the cell lines, suggesting antioxidant activity. Of the 84 antioxidant pathway genes included in microarray array analysis (HT-29 cells), the expressions of 28 genes were enhanced (P<0.05). Our findings suggest that carotenoids of germ, aleurone, and endosperm fractions improved antioxidant capacity and thus have the potential to mitigate oxidative stress.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)