Merck
Particle size influences fibronectin internalization and degradation by fibroblasts.
Experimental cell research (2014-07-06)
Peter Bozavikov, Dhaarmini Rajshankar, Wilson Lee, Christopher A McCulloch
ZUSAMMENFASSUNG
The application of nanotechnology for drug targeting underlines the importance of controlling the kinetics and cellular sites of delivery for optimal therapeutic outcomes. Here we examined the effect of particle size on internalization and degradation of surface-bound fibronectin by fibroblasts using polystyrene nanoparticles (NPs; 51 nm) and microparticles (MPs; 1 μm). Fibronectin was strongly bound by NPs and MPs as assessed by immuno-dot blot analysis (5.1 ± 0.4 × 10(- 5)pg fibronectin per μm(2) of NP surface; 4.2 ± ± 0.3 × 10(-5)pg fibronectin per μm(2) of MP surface; p>0.2). We estimated that ~193 fibronectin molecules bound to a MP compared with 0.6 fibronectin molecules per NP, indicating that ~40% of nanoparticles were not bound by fibronectin. One hour after incubation, fibronectin-coated NPs and MPs were rapidly internalized by Rat-2 fibroblasts. MPs and NPs were engulfed partly by receptor-mediated endocytosis as indicated by decreased uptake when incubated at 4°C, or by depletion of ATP with sodium azide. Pulse-chase experiments showed minimal exocytosis of NPs and MPs. Internalization of NPs and MPs was inhibited by jasplakinolide, whereas internalization of MPs but not NPs was inhibited by latrunculin B and by integrin-blocking antibodies. Extraction of plasma membrane cholesterol with methyl β-cyclodextrin inhibited internalization of fibronectin-coated NPs but not MPs. Biotinylated fibronectin internalized by cells was extensively degraded on MPs but not NPs. Particle size affects actin and clathrin-dependent internalization mechanisms leading to fibronectin degradation on MPs but not NPs. Thus either prolonged, controlled release or an immediate delivery of drugs can be achieved by adjusting the particle size along with matrix proteins such as FN.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Chlorpromazin -hydrochlorid, ≥98% (TLC)
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Supelco
Genistein, analytical standard
USP
Chlorpromazin -hydrochlorid, United States Pharmacopeia (USP) Reference Standard
Supelco
Chlorpromazin -hydrochlorid, VETRANAL®, analytical standard
Sigma-Aldrich
Chlorpromazin -hydrochlorid, meets USP testing specifications
Chlorpromazin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard