Merck
  • Optimization and validation of a chiral GC-MS method for the determination of free D-amino acids ratio in human urine: application to a gestational diabetes mellitus study.
Optimization and validation of a chiral GC-MS method for the determination of free D-amino acids ratio in human urine: application to a gestational diabetes mellitus study.
Journal of pharmaceutical and biomedical analysis (2015-02-14)
Ma Paz Lorenzo, Danuta Dudzik, Elena Varas, Manuel Gibellini, Mariusz Skotnicki, Marcin Zorawski, Wieslaw Zarzycki, Federica Pellati, Antonia García
ZUSAMMENFASSUNG
Gestational Diabetes Mellitus (GDM) is defined as glucose intolerance with onset or first recognition during pregnancy. It is affecting approximately up to 14% of all pregnancies with an increasing tendency. GDM has been related to relevant short-term and long-term health complications for both mother and offspring. Recent studies strongly emphasized the role of several essential amino acids in the pathogenesis of obesity and highlighted their strong correlation with insulin resistance, but there are no references related to modifications in D-AAs in biological fluids. As D-AA elimination proceeds mainly by renal excretion, urine was the selected sample to evaluate the alterations in free D-AAs ratio in a GDM study. Only 1 mL of first void urine or standard solution was required for purification, by using a Discovery DSC-SCX SPE cartridge (500 mg/3 mL) and derivatization into their N(O)-pentafluoropropionyl amino acid 2-propyl esters. Enantiomeric separation was carried out by GC-MS on a Chirasil-L-Val N-propionyl-L-valine-tert-butylamide polysiloxane fused-silica capillary column (25 m×0.25 mm I.D., 0.12 μm film thickness, Agilent Technologies, Waldbronn, Germany), under programmed temperature elution. Detection was performed with an ion trap mass analyzer, operating in the full scan mode in the m/z 50-350 range. 14 pairs of derivatives of D-and L-AAs were separated. The steps of sample preparation, derivatization and GC-MS conditions were optimized for both urine and standards. Several conditions affecting the SPE procedure, such as sorbent mass/volume ratio of the cartridge, sample dilution and pH, were optimized. Volume of reagents and solvents and reaction temperature and time were also tested for the derivatization. Regarding the GC-MS parameters, split ratio, temperature program and mass range were optimized. The final method was validated in terms of linearity, sensitivity, accuracy and precision for D-Ala, D-Pro, D-Ser, D-Met, D-Phe, D-Glu, D-Orn and D-Lys. Identification of AAs in urine samples was based on retention time and mass spectra. Urine from 20 women with GDM and 20 pregnant women with normal glucose tolerance (after 2-h 75-g oral glucose tolerance test), matched according to the week of gestation and age (22-28 week of gestation and age 24-37 years), were enrolled into the study. %D-Relative amounts were determined for Ala, Val, Thr, Ser, Leu, Asx (Asp+Asn), Glx (Glu+Gln), Met, Phe, Tyr, Orn and Lys. Statistically significant differences (p<0.05) were observed only for D-Phe and higher values were found in the GDM group. It is possible that D-Phe could be involved in metabolic/signaling pathways to compensate early stages of insulin resistance, although further work is necessary to confirm this hypothesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, ≥99.0% (GC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Methionin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Isoleucin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Butyliertes Hydroxytoluol, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Threonin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Butyliertes Hydroxytoluol, ≥99%, FCC, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.