Merck
  • The short-chain fatty acid receptor GPR43 is transcriptionally regulated by XBP1 in human monocytes.
The short-chain fatty acid receptor GPR43 is transcriptionally regulated by XBP1 in human monocytes.
Scientific reports (2015-01-31)
Zhiwei Ang, Jun Zhi Er, Jeak Ling Ding
ZUSAMMENFASSUNG
G-protein coupled receptor 43 (GPR43) recognizes short chain fatty acids and is implicated in obesity, colitis, asthma and arthritis. Here, we present the first full characterization of the GPR43 promoter and 5'-UTR. 5'-RACE of the GPR43 transcript identified the transcription start site (TSS) and a 124 bp 5'-UTR followed by a 1335 bp intron upstream of the ATG start codon. The sequence spanning -4560 to +68 bp relative to the GPR43 TSS was found to contain strong promoter activity, increasing luciferase reporter expression by >100-fold in U937 monocytes. Stepwise deletions further narrowed the putative GPR43 promoter (-451 to +68). Site-directed mutagenesis identified XBP1 as a core cis element, the mutation of which abrogated transcriptional activity. Mutations of predicted CREB, CHOP, NFAT and STAT5 binding sites, partially reduced promoter activity. ChIP assays confirmed the binding of XBP1 to the endogenous GPR43 promoter. Consistently, GPR43 expression is reduced in monocytes upon siRNA-knockdown of XBP1, while A549 cells overexpressing XBP1 displayed elevated GPR43 levels. Based on its ability to activate XBP1, we predicted and confirmed that TNFα induces GPR43 expression in human monocytes. Altogether, our findings form the basis for strategic modulation of GPR43 expression, with a view to regulate GPR43-associated diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Natriumacetat, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Natriumacetat, ACS reagent, ≥99.0%
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Natriumacetat, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Anti-Aktin, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Phenol -Lösung, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Verflüssigtes Phenol, ≥89.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Natriumacetat, 99.995% trace metals basis
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumacetat, >99%, FG
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Phenol, Molecular Biology