Merck
  • Comparison of a stable isotope labeled (SIL) peptide and an extended SIL peptide as internal standards to track digestion variability of an unstable signature peptide during quantification of a cancer biomarker, human osteopontin, from plasma using capillary microflow LC-MS/MS.
Comparison of a stable isotope labeled (SIL) peptide and an extended SIL peptide as internal standards to track digestion variability of an unstable signature peptide during quantification of a cancer biomarker, human osteopontin, from plasma using capillary microflow LC-MS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-08-19)
Morse Faria, Matthew S Halquist, Moucun Yuan, William Mylott, Rand G Jenkins, H Thomas Karnes
ZUSAMMENFASSUNG
Human osteopontin (hOPN) is a secreted plasma protein which is elevated in various cancers and is indicative of poor prognosis. Here we describe investigations involving an extended peptide internal standard to track an unstable signature peptide followed by further method development and validation for quantitative measurement of hOPN from plasma using microflow liquid chromatography and tandem mass spectrometry (MFLC-MS/MS). A biologically relevant tryptic peptide 'GDSVVYGLR' was used as a signature peptide for this method. The optimized method involved immunocapture of the analyte protein using hOPN specific antibodies followed by trypsin digestion to obtain the signature peptide. Analysis was carried out on a Waters IonKey/MS system using a flow rate of 2.5μL/min. Immunocapture buffer was used as a surrogate matrix for the validation studies. The method was validated over a range of 25-600ng/mL. Intra-assay and inter-assay accuracies were within ±13%. Intra-assay and inter-assay precision were within 17%. A stable isotope labeled (SIL) peptide GDSVVYGLR* and an extended SIL peptide TYDGRGDSVV*YGLRSKSKKF were evaluated as internal standards (IS) to account for signature peptide digestion instability and variability. Inherent digestion variability i.e., under controlled conditions, was within ±20% with both IS peptides. In digestion variability studies, where trypsin activity was varied (20-180%), the use of the extended SIL peptide as an internal standard limited the variability to within ±30% of the normalized response. Alternatively, when the SIL peptide was used as the internal standard, the variability ranged from -67.4% to 50.6% of the normalized response. The applicability of the validated method was demonstrated by quantification of OPN from plasma samples obtained from 10 healthy individuals and 10 breast cancer patients. The plasma OPN concentrations in healthy individuals ranged from 38 to 85ng/mL with a mean concentration of 55.4±15.3ng/mL. A 1.5-12 fold increase in OPN concentrations, ranging from 85 to 637ng/mL, was seen in breast cancer patient samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
Natriumchlorid -Lösung, 5 M
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%