Merck
  • Antioxidant capacity and chemical profiles of Satureja montana L. Honey: hotrienol and syringyl derivatives as biomarkers.
Antioxidant capacity and chemical profiles of Satureja montana L. Honey: hotrienol and syringyl derivatives as biomarkers.
Chemistry & biodiversity (2015-07-15)
Igor Jerković, Carlo I G Tuberoso, Zvonimir Marijanović, Marina Kranjac, M Malenica-Staver
ZUSAMMENFASSUNG
The present study is focused on the antioxidant capacity and chemical profiling of eight Croatian Satureja montana L. honey samples. Among the 20 compounds obtained by headspace solid-phase microextraction (HS-SPME) and identified by GC-FID and GC/MS analyses, hotrienol was predominant (75.9-81.7%). The honey matrix volatile/semivolatile profile was investigated by ultrasonic solvent extraction (USE) followed by GC-FID and GC/MS analyses. The major compounds identified by this latter method were the sinapic-acid derivatives methyl syringate (36.2-72.8%) and syringaldehyde (2.2-43.1%). Direct, targeted HPLC-DAD analyses of the native honey samples revealed the presence of methyl syringate (7.10-39.60 mg/kg) and syringic acid (0.10-1.70 mg/kg). In addition, the total phenolic content of the samples was determined by the FolinCiocalteu assay (311.0-465.9 mg GAE/kg), and the antioxidant capacity was evaluated by the DPPH radical-scavenging activity (0.5-1.0 mmol TEAC/kg) and the ferric reducing antioxidant power (2.5-5.1 mmol Fe(2+) /kg).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchroman-2-carbonsäure, 97%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
SAFC
Natriumchlorid -Lösung, 5 M
Supelco
2,4,6-Tri-(2-pyridyl)-s-triazin, ≥98%
Sigma-Aldrich
Divinylbenzol, technical grade, 55%
Sigma-Aldrich
Divinylbenzol, technical grade, 80%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumcarbonat, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Natriumcarbonat, BioXtra, ≥99.0%
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Wasser, BioPerformance Certified
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Sigma-Aldrich
Poly(dimethylsiloxan), viscosity 1.0 cSt (25 °C)
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub