Merck
Novel Dammarane-Type Triterpene Saponins from Panax ginseng Root.
Chemical & pharmaceutical bulletin (2015-11-03)
Dong Gu Lee, Ah Young Lee, Kyung-Tack Kim, Eun Ju Cho, Sanghyun Lee
ZUSAMMENFASSUNG
Four phytochemical constituents were isolated from Panax ginseng root by repeated column chromatography (CC), medium pressure liquid chromatography (MPLC), high-speed counter current chromatography (HSCCC), and semi-preparative HPLC. Their structures were elucidated as the dammarane-type triterpene saponins ginsenoside-Rg18 (1), 6-acetyl ginsenoside-Rg3 (2), ginsenoside-Rs11 (3), and ginsenoside-Re7 (4) based on spectral data. Compounds 1-4 from P. ginseng root were new compounds from nature. They showed good hydroxyl radical scavenging activity and anti-bacterial activity against Escherichia coli and Staphylococcus aureus. However, they did not show any anti-inflammatory activity. In addition, they inhibited the growth of adenocarcinoma gastric stomach cells. Among them, ginsenoside-Rs11 (3) showed the best anti-oxidative, anti-bacterial, and anti-cancer activities.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Schwefelsäure, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbisulfat
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Tetramethylsilan, NMR grade, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
2-Desoxy-D-ribose, 97% (GC)