Merck
  • C-Terminally Truncated Forms of Tau, But Not Full-Length Tau or Its C-Terminal Fragments, Are Released from Neurons Independently of Cell Death.
C-Terminally Truncated Forms of Tau, But Not Full-Length Tau or Its C-Terminal Fragments, Are Released from Neurons Independently of Cell Death.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2015-08-01)
Daniel Kanmert, Adam Cantlon, Christina R Muratore, Ming Jin, Tiernan T O'Malley, Gloria Lee, Tracy L Young-Pearse, Dennis J Selkoe, Dominic M Walsh
ZUSAMMENFASSUNG
Recent evidence suggests that tau aggregation may spread via extracellular release and subsequent uptake by synaptically connected neurons, but little is known about the processes by which tau is released or the molecular forms of extracellular tau. To gain insight into the nature of extracellular tau, we used highly sensitive ELISAs, which, when used in tandem, are capable of differentiating between full-length (FL) tau, mid-region-bearing fragments, and C-terminal (CT) fragments. We applied these assays to the systematic study of the conditioned media of N2a cells, induced pluripotent stem cell-derived human cortical neurons, and primary rat cortical neurons, each of which was carefully assessed for viability. In all three neuronal models, the bulk of extracellular tau was free-floating and unaggregated and <0.2% was encapsulated in exosomes. Although most intracellular tau was FL, the majority of extracellular tau was CT truncated and appeared to be released both actively by living neurons and passively by dead cells. In contrast, only a small amount of extracellular tau was aggregation-competent tau (i.e., contained the microtubule-binding regions) and this material appears to be released solely due to a low level of cell death that occurs in all cell culture systems. Importantly, amyloid β-protein (Aβ)-induced neuronal compromise significantly increased the quantity of all forms of extracellular tau, but the presence of Aβ before detectable cell compromise did not increase extracellular tau. Collectively, these results suggest that factors that induce neuronal death are likely to be necessary to initiate the extracellular spread of tau aggregation. Recent studies suggest that the transfer of tau between neurons underlies the characteristic spatiotemporal progression of neurofibrillary pathology. We searched for tau in the conditioned medium of N2a cells, induced pluripotent stem cell-derived human cortical neurons, and primary rat cortical neurons and analyzed the material present using four different tau ELISAs. We demonstrate that the majority of tau released from healthy neurons is C-terminally truncated and lacks the microtubule-binding region (MTBR) thought necessary for self-aggregation. A small amount of MTBR-containing tau is present outside of cells, but this appears to be solely due to cell death. Therefore, if propagation of tau aggregation is mediated by extracellular tau, our findings suggest that neuronal compromise is required to facilitate this process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
1,1,1,3,3,3-Hexafluor-2-propanol, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Aluminiumoxid, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Aluminiumoxid, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Aluminiumoxid, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)