Merck
  • Ring substitution influences oxidative cyclisation and reactive metabolite formation of nordihydroguaiaretic acid analogues.
Ring substitution influences oxidative cyclisation and reactive metabolite formation of nordihydroguaiaretic acid analogues.
Bioorganic & medicinal chemistry (2015-10-07)
Isaac Asiamah, Heather L Hodgson, Katherine Maloney, Kevin J H Allen, Ed S Krol
ZUSAMMENFASSUNG
Nordihydroguaiaretic acid (NDGA) is a natural polyphenol with a broad spectrum of pharmacological properties. However, its usefulness is hindered by the lack of understanding of its pharmacological and toxicological pathways. Previously we showed that oxidative cyclisation of NDGA at physiological pH forms a dibenzocyclooctadiene that may have therapeutic benefits whilst oxidation to an ortho-quinone likely mediates toxicological properties. NDGA analogues with higher propensity to cyclise under physiologically relevant conditions might have pharmacological implications, which motivated this study. We synthesized a series of NDGA analogues which were designed to investigate the structural features which influence the intramolecular cyclisation process and help to understand the mechanism of NDGA's autoxidative conversion to a dibenzocyclooctadiene lignan. We determined the ability of the NDGA analogues investigated to form dibenzocyclooctadienes and evaluated the oxidative stability at pH 7.4 of the analogues and the stability of any dibenzocyclooctadienes formed from the NDGA analogues. We found among our group of analogues the catechols were less stable than phenols, a single catechol-substituted ring is insufficient to form a dibenzocyclooctadiene lignan, and only compounds possessing a di-catechol could form dibenzocyclooctadienes. This suggests that quinone formation may not be necessary for cyclisation to occur and the intramolecular cyclisation likely involves a radical-mediated rather than an electrophilic substitution process. We also determined that the catechol dibenzocyclooctadienes autoxidised at comparable rates to the parent catechol. This suggests that assigning in vitro biological activity to the NDGA dibenzocyclooctadiene is premature and requires additional study.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Silbernitrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥98%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Magnesiumsulfat, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Silbernitrat, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Zitronensäure, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Magnesiumsulfat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
Magnesiumsulfat, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Silbernitrat, 99.9999% trace metals basis
Sigma-Aldrich
Zitronensäure, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Magnesiumsulfat, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Kaliumphosphat, anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Kaliumphosphat, reagent grade, ≥98.0%
Sigma-Aldrich
Magnesiumsulfat, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesiumsulfat -Lösung, Molecular Biology, 1.00 M±0.04 M