Merck
  • Intravenous Preload of Mesenchymal Stem Cells Rescues Erectile Function in a Rat Model of Cavernous Nerve Injury.
Intravenous Preload of Mesenchymal Stem Cells Rescues Erectile Function in a Rat Model of Cavernous Nerve Injury.
The journal of sexual medicine (2015-07-28)
Akio Takayanagi, Masanori Sasaki, Yuko Kataoka-Sasaki, Ko Kobayashi, Yohei Matsuda, Shinichi Oka, Naoya Masumori, Jeffery D Kocsis, Osamu Honmou
ZUSAMMENFASSUNG
We evaluated the potential preventive effects and mechanisms of intravenously preloaded mesenchymal stem cells (MSCs) for erectile dysfunction (ED) in a cavernous nerve (CN) injury model. Male Sprague-Dawley (SD) rats were used for this study. Rats were randomized into two groups. One group was intravenously preloaded with MSCs (1.0 × 10(6) cells in 1 mL total fluid volume) and the other was infused with medium alone (1 mL Dulbecco's modified Eagle's medium [DMEM]) for sham control, respectively. Crushed CN injury was induced immediately after infusion. The surgeon was blind to the experimental conditions (MSC or medium). To assess erectile function, we measured the intracavernous pressure (ICP) and arterial pressure (AP) at 1 hour and 2 weeks after CN injury. After measuring the initial ICP/AP of pre-injury (normal) male SD rats, they were randomized into the two groups and infused with MSCs or medium. PKH26-labelled MSCs were used for tracking. To investigate the mRNA expression levels of neurotrophins in the major pelvic ganglia (MPG), we performed real-time quantitative real-time polymerase chain reaction. The reduction of ICP/AP and area under the curve of ICP (ICP-AUC) in the MSC group was significantly lower than in the DMEM group (P < 0.05; P < 0.05) at 1 hour. The ICP/AP and ICP-AUC at 2 weeks post-injury in the MSC group was significantly higher than in the DMEM group (P < 0.01; P < 0.05). The preloaded PKH26-labelled MSCs were detected in the MPG and CN using confocal microscopy indicating homing of the cells to the injured nerve and ganglia. Glia cell-derived neurotrophic factor (GDNF) and neurturin, which are important neurotrophic factors for erection, had expression levels in MPG significantly higher in the MSC group than in the DMEM group (P < 0.01, 0.05). Intravenous preload of MSCs before a CN injury may prevent or reduce experimental ED.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
L-Glutamin
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Glutamin
Preise und Verfügbarkeit sind derzeit nicht verfügbar.