Merck
SIRT3 deficiency impairs mitochondrial and contractile function in the heart.
Basic research in cardiology (2015-05-13)
Christoph Koentges, Katharina Pfeil, Tilman Schnick, Sebastian Wiese, Rabea Dahlbock, Maria C Cimolai, Maximilian Meyer-Steenbuck, Katarina Cenkerova, Michael M Hoffmann, Carsten Jaeger, Katja E Odening, Bernd Kammerer, Lutz Hein, Christoph Bode, Heiko Bugger
ZUSAMMENFASSUNG
Sirtuin 3 (SIRT3) is a mitochondrial NAD(+)-dependent deacetylase that regulates energy metabolic enzymes by reversible protein lysine acetylation in various extracardiac tissues. The role of SIRT3 in myocardial energetics and in the development of mitochondrial dysfunction in cardiac pathologies, such as the failing heart, remains to be elucidated. To investigate the role of SIRT3 in the regulation of myocardial energetics and function SIRT3(-/-) mice developed progressive age-related deterioration of cardiac function, as evidenced by a decrease in ejection fraction and an increase in enddiastolic volume at 24 but not 8 weeks of age using echocardiography. Four weeks following transverse aortic constriction, ejection fraction was further decreased in SIRT3(-/-) mice compared to WT mice, accompanied by a greater degree of cardiac hypertrophy and fibrosis. In isolated working hearts, a decrease in cardiac function in SIRT3(-/-) mice was accompanied by a decrease in palmitate oxidation, glucose oxidation, and oxygen consumption, whereas rates of glycolysis were increased. Respiratory capacity and ATP synthesis were decreased in cardiac mitochondria of SIRT3(-/-) mice. HPLC measurements revealed a decrease of the myocardial ATP/AMP ratio and of myocardial energy charge. Using LC-MS/MS, we identified increased acetylation of 84 mitochondrial proteins, including 6 enzymes of fatty acid import and oxidation, 50 subunits of the electron transport chain, and 3 enzymes of the tricarboxylic acid cycle. Lack of SIRT3 impairs mitochondrial and contractile function in the heart, likely due to increased acetylation of various energy metabolic proteins and subsequent myocardial energy depletion.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥95% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥99%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Adonitol, ≥99%
Sigma-Aldrich
Xanthin, ≥99%
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast