Merck
  • Identification of Novel Components Influencing Colonization Factor Antigen I Expression in Enterotoxigenic Escherichia coli.
Identification of Novel Components Influencing Colonization Factor Antigen I Expression in Enterotoxigenic Escherichia coli.
PloS one (2015-10-31)
Sara Haines, Sylviane Gautheron, William Nasser, Geneviève Renauld-Mongénie
ZUSAMMENFASSUNG
Colonization factors (CFs) mediate early adhesion of Enterotoxigenic Escherichia coli (ETEC) in the small intestine. Environmental signals including bile, glucose, and contact with epithelial cells have previously been shown to modulate CF expression in a strain dependent manner. To identify novel components modulating CF surface expression, 20 components relevant to the intestinal environment were selected for evaluation. These included mucin, bicarbonate, norepinephrine, lincomycin, carbon sources, and cations. Effects of individual components on surface expression of the archetype CF, CFA/I, were screened using a fractional factorial Hadamard matrix incorporating 24 growth conditions. As most CFs agglutinate erythrocytes, surface expression was evaluated by mannose resistant hemagglutination. Seven components, including porcine gastric mucin, lincomycin, glutamine, and glucose were found to induce CFA/I surface expression in vitro in a minimal media while five others were inhibitory, including leucine and 1,10-phenanthroline. To further explore the effect of components positively influencing CFA/I surface expression, a response surface methodology (RSM) was designed incorporating 36 growth conditions. The optimum concentration for each component was identified, thereby generating a novel culture media, SP1, for CFA/I expression. CFs closely related to CFA/I, including CS4 and CS14 were similarly induced in SP1 media. Other epidemiologically relevant CFs were also induced when compared to the level obtained in minimal media. These results indicate that although CF surface expression is complex and highly variable among strains, the CF response can be predicted for closely related strains. A novel culture media inducing CFs in the CF5a group was successfully identified. In addition, mucin was found to positively influence CF expression in strains expressing either CFA/I or CS1 and CS3, and may function as a common environmental cue.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Asparaginsäure, reagent grade, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ammoniumchlorid, ReagentPlus®, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycerin, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Lysin, ≥98% (TLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC), BioXtra
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Leucin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Methionin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
1,10-Phenanthrolin, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ammoniumchlorid, Molecular Biology, suitable for cell culture, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-Leucin, reagent grade, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
L-Glutamin
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.