Merck
Catalytic metallodrugs based on the LaR2C peptide target HCV SLIV IRES RNA.
Dalton transactions (Cambridge, England : 2003) (2015-11-20)
Martin James Ross, Seth S Bradford, J A Cowan
ZUSAMMENFASSUNG
Prior work has demonstrated the potential effectiveness of a new class of metallopeptides as catalytic metallodrugs that target HCV IRES SLIIb RNA (Cu-GGHYrFK, 1). Herein new catalytic metallodrugs (GGHKYKETDLLILFKDDYFAKKNEERK, 2; and GGHKYKETDL, 3) are described based on the LaR2C peptide that has been shown to bind to the SLIV HCV IRES domain. In vitro fluorescence assays yielded KD values ∼10 μM for both peptides and reaction of the copper derivatives with SLIV RNA demonstrated initial rates comparable across different assays as well as displaying pseudo-Michaelis-Menten behavior. The sites of reaction and cleavage mechanisms were determined by MALDI-TOF mass spectrometry. The primary site of copper-promoted SLIV cleavage is shown to occur in the vicinity of the 5'-G17C18A19C20-3' sequence that corresponds to a known binding site of the RM2 motif of the human La protein and has previously been reported to be important for viral translation. This domain also flanks the internal start codon (AUG). Both copper complexes also showed efficacy in an HCV replicon assay (IC50 = 0.75 μM for 2-Cu, and 2.17 μM for 3-Cu) and show potential for treatment of hepatitis C, complementing other marketed drugs by acting on a distinct therapeutic target by a novel mechanism of action.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Formamid, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Formamid, ≥99.5% (GC), BioReagent, Molecular Biology
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Formamid, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
SAFC
Natriumchlorid -Lösung, 5 M